Adapters for TX-150 Swinging Bucket Rotors
Adapters for TX-150 Swinging Bucket Rotors
Thermo Scientificā„¢
Adapters for TX-150 Swinging Bucket Rotors
Assorted tube adapters
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTube Type
750057395/7 mL Blood Collection Tube
7500574425 mL Universal Flat/Skirted Tube
750057403/5 mL Blood Collection Tube
7500573711 mL IVF Tube
7500573650 mL Conical or Skirted Tube
75005735100 mL Round Bottom Open-Top Tube
75005743Microtube
7500350413 mL Urine Tube
Catalog number 75005739
Price (USD)/ Pack of 4
501.30
Online exclusive
557.00 
Save 55.70 (10%)
-
Add to cart
Tube Type:
5/7 mL Blood Collection Tube
Price (USD)/ Pack of 4
501.30
Online exclusive
557.00 
Save 55.70 (10%)
Add to cart
Adapters for TX-150 Swinging Bucket Rotors
Catalog number75005739
Price (USD)/ Pack of 4
501.30
Online exclusive
557.00 
Save 55.70 (10%)
-
Add to cart
Assorted tube adapters
Specifications
TypeRotor Adapter
Tube Type5/7 mL Blood Collection Tube
Capacity5/7 mL
Max. Tube Diameter13 mm
Max. Tube Length116 mm
For Use With (Equipment)50mL Conical Buckets for TX-150 rotor
Unit SizePack of 4