Search Thermo Fisher Scientific
Catalog Number | Description |
---|---|
75006478 | DoubleSpin Bucket |
75006421 | Sealing Cap |
75006436 | Double Blood Bag Bucket |
75006476 | LH-4000W Windshield Swinging Bucket Rotor |
75006477 | Round Bucket |
75006479 | Biocontainment Sealing Lid |
75006486 | Microplate Adapter |
Support a diverse array of sample processing applications without the need to modify existing protocols with the Thermo Scientific™ LH-4000W 8 x 500mL Windshield Swinging Bucket Rotor. Choose from round, DoubleSpin™ and Double blood bag buckets with a wide array of available adapters, providing capacity flexibility ranging from 1.5 to 250mL.
Buckets, biocontainment lids and DoubleSpin buckets are sold separately; order rotor body (75006476) and components to complete assembly. Tube adapters and microplate adapter for DoubleSpin bucket are also available separately. Contact your Sales Representative for information.
Compatible with:
Thermo Scientific™ Cryofuge 5500i series centrifuges