Sorvall ST1 R Plus, 120V TX-400 Cell Culture Package
Sorvall ST1 R Plus, 120V TX-400 Cell Culture Package
Thermo Scientific™
Sorvall ST1 R Plus, 120V TX-400 Cell Culture Package
Green features
Waste no time finding the right part numbers for your application! The application driven ce ntrifuge package ensures convenient, fast ordering of all components necessary to run a specific protocol.
Have Questions?
Número de catálogo 75016014
Precio (EUR)
-
Solicite un presupuesto
Ask our AI about this Product
Waste no time finding the right part numbers for your application! The application driven ce ntrifuge package ensures convenient, fast ordering of all components necessary to run a specific protocol. Run 16 x 50 mL conicals under biocontainment certified lids. Contains centrifuge, TX-400 rotor, buckets, biocontainment lids, 50 mL conical adapters (maximum capacity 16 x 50mL), 15 mL conical adapter (maximum capacity 36 x 15 mL).
  • Easy to Use:
    • Intuitive controls simplify operation and provide detailed information on a wide range of operating parameters and processing status for optimal performance.
    • Modern industrial design supports continuous improvement in rotor loading ergonomics, user safety and current needed certifications
    • Motorized lid latch enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches.
    Thermo Scientific™ Auto-Lock™ III Rotor System:
    • Secure locking system allows easy push-button installation and exchange of rotors.
    • System saves time when switching between applications.
    • Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life.
    Thermo Scientific™ ClickSeal™ Bucket Sealing System:
    • Biocontainment solution seals with a snap; eliminates screw caps and complicated clips.
    • System has glove-friendly, one-handed open/close capability.
    • System is certified by CAMR™ in Porton Down, UK.
    Thermo Scientific™ SMARTSpin™ Technology:
    • Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option:
    • Technology optimizes safety and improves separations.
    Space-Saving Design:
    • Compact dimensions optimize work area and ergonomic height and curved design ensure easy sample loading and unloading and easy cleaning.
    Energy Efficient:
    • System offers up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing.
Especificaciones
Sistema de accionamientoDirect, Brushless induction low profile motor with choices of 9 acceleration, 10 deceleration rates
Características ecológicasLess waste, Energy efficient
IncluyePart Numbers In Pk. 75009381 (centrifuge),75003181(rotor),75003655(set of buckets),75003656(lids),75003682(set of adapters),75003683(set of adapters)
TipoSerie de centrífugas de uso general
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documentos y descargas
Certificados
Se muestran 0 resultados, busque arriba un certificado específico
Compartir número de catálogo, nombre o enlace.
1x1 image pixel for data collection