Thermo Scientific™ Multifuge™ X4 and X4R General Purpose Pro Centrifuges provide high performance and versatility with an easy-to-use, full-color touch screen user interface. The centrifuges make programming easy for quick results and allow easy access to and tracking of all runs, rotor life and centrifuge health. With up to 4L capacity, they allow spinning 196 blood tubes, 40x 50mL conical tubes and 96x 15mL conical tubes. Maximum versatility is achieved with 18 available rotors to choose from and Thermo Scientific™ Auto-Lock™ Rotor Exchange to quickly change between applications.
Available for purchase individually or in convenient application-based packages: Cell Culture packages include the centrifuge, rotor, ClickSeal biocontainment lids and a set of four 15 mL and 50 mL conical tube adapters.
Blood Tube packages include the centrifuge, rotor, ClickSeal biocontainment lids and a set of four 5/7 mL and 9/10 mL blood tube adapters.
196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes
Max. RCF
4,122 x g (with TX-1000 rotor)
Max. Speed
4,200 rpm (with TX-1000 rotor)
Product Line
Multifuge
Refrigerated
Yes
Display
Full Color Touch Screen Interface
Drive System
Direct, Brushless Induction Low Profile
Green Features
Energy efficient, less waste
Includes
Printed English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Memory
Stores up to 100 events and programs
Profile (Acceleration/Braking)
Choices of 9 Accel/10 Decel Rates
Program Storage
Up to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor
Blood Tube Package Includes TX-1000 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube Adapters
Rotor Included
Yes
Run Time
Up to 99 hr. 59 min. 59 sec.
Standards
ISO 14971, ISO 13485, ISO 9001
Temperature Range
-10°C to +40°C, ±2°C
Timer
Option to count up or down and to set “At start” or “At speed”
Type
Benchtop Centrifuge
Ventilated
No
Warranty
Centrifuge: 5 Year Limited, Rotor: See Manual
Net Weight
See individual components of package
Voltage
120 V
Frequency
60 Hz
Laboratory/Cleanroom Use
Cleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller Type
Microprocessor
Description
Multifuge X4 Pro-MD 208-240V 50/60Hz
Capacity
4 x 1000 mL BioBottles, 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 4 x 6 Microplates with TX-1000 Rotor
Max. RCF
25,830 x g Fixed Angle with Microliter 30 x 2 Rotor, 7,197 x g Swing-Out with H-Flex HS4 Rotor
Product Line
Multifuge
Refrigerated
No
Display
Full Color Touch Screen Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
Printed English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Memory
Stores up to 100 events and programs
Model
IVD Certified
Noise Level
<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)
Choices of 9 Accel/10 Decel Rates
Program Storage
Up to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor
18 Rotor options available
Rotor Included
No
Run Time
Up to 99 hr. 59 min. 59 sec.
Standards
ISO 14971, ISO 13485, ISO 9001
Timer
Option to count up or down and to set 'At start' or 'At speed'
Type
Benchtop Centrifuge
Ventilated
Yes
Warranty
5 Year Limited
Dimensions (L x W x H) Exterior
690 x 566 x 362 mm
Length (English) Exterior
27.2 in.
Length (Metric) Exterior
690 mm
Width (English) Exterior
22.3 in.
Width (Metric) Exterior
566 mm
Height (English) Exterior
14.2 in.
Height (English) Exterior Lid Open
33.9 in.
Height (Metric) Exterior
362 mm
Height (Metric) Exterior Lid Open
861 mm
Depth (English)
27.2 in.
Depth (Metric)
690 mm
Net Weight
89 kg
Voltage
208/240 V
Wattage
1600 w
Frequency
50/60 Hz
Laboratory/Cleanroom Use
Cleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller Type
Microprocessor
Description
Multifuge X4 Pro-MD 120V
Capacity
4 x 1000 mL BioBottles, 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 4 x 6 Microplates with TX-1000 Rotor
Max. RCF
25,830 x g Fixed Angle with Microliter 30 x 2 Rotor, 7,197 x g Swing-Out with H-Flex HS4 Rotor
Product Line
Multifuge
Refrigerated
No
Display
Full Color Touch Screen Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
Printed English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Memory
Stores up to 100 events and programs
Model
IVD Certified
Noise Level
<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)
Choices of 9 Accel/10 Decel Rates
Program Storage
Up to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor
18 Rotor options available
Rotor Included
No
Run Time
Up to 99 hr. 59 min. 59 sec.
Standards
ISO 14971, ISO 13485, ISO 9001
Timer
Option to count up or down and to set 'At start' or 'At speed'
Type
Benchtop Centrifuge
Ventilated
Yes
Warranty
5 Year Limited
Dimensions (L x W x H) Exterior
690 x 566 x 362 mm
Length (English) Exterior
27.2 in.
Length (Metric) Exterior
690 mm
Width (English) Exterior
22.3 in.
Width (Metric) Exterior
566 mm
Height (English) Exterior
14.2 in.
Height (English) Exterior Lid Open
33.9 in.
Height (Metric) Exterior
362 mm
Height (Metric) Exterior Lid Open
861 mm
Depth (English)
27.2 in.
Depth (Metric)
690 mm
Net Weight
89 kg
Voltage
120 V
Wattage
1300 w
Frequency
50/60 Hz
Laboratory/Cleanroom Use
Cleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller Type
Microprocessor
Description
Multifuge X4R Pro-MD 220-240V 50Hz / 230V 60Hz
Capacity
4 x 1000 mL BioBottles, 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 4 x 6 Microplates with TX-1000 Rotor
Max. RCF
25,830 x g Fixed Angle with Microliter 30 x 2 rotor, 7,197 x g Swing-Out with H-Flex HS4 rotor
Product Line
Multifuge
Refrigerated
Yes
Display
Full Color Touch Screen Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
Printed English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Memory
Stores up to 100 events and programs
Model
IVD Certified
Noise Level
<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)
Choices of 9 Accel/10 Decel Rates
Program Storage
Up to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor
18 Rotor options available
Rotor Included
No
Run Time
Up to 99 hr. 59 min. 59 sec.
Standards
ISO 14971, ISO 13485, ISO 9001
Temperature Range
-10°C to +40°C, ±2°C
Timer
Option to count up or down and to set “At start” or “At speed”
Type
Benchtop Centrifuge
Ventilated
No
Warranty
5 Year Limited
Dimensions (L x W x H) Exterior
690 x 746 x 361 mm
Length (English) Exterior
27.2 in.
Length (Metric) Exterior
690 mm
Width (English) Exterior
29.4 in.
Width (Metric) Exterior
746 mm
Height (English) Exterior
14.2 in.
Height (English) Exterior Lid Open
33.9 in.
Height (Metric) Exterior
361 mm
Height (Metric) Exterior Lid Open
861 mm
Depth (English)
27.2 in.
Depth (Metric)
690 mm
Net Weight
120 kg
Voltage
220-240 V
Wattage
1950 w
Frequency
50/60 Hz
Laboratory/Cleanroom Use
Cleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller Type
Microprocessor
Description
Multifuge X4R Pro-MD 120V
Capacity
4 x 1000 mL BioBottles, 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 4 x 6 Microplates with TX-1000 Rotor
Max. RCF
25,830 x g Fixed Angle with Microliter 30 x 2 rotor, 7,197 x g Swing-Out with H-Flex HS4 rotor
Product Line
Multifuge
Refrigerated
Yes
Display
Full Color Touch Screen Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
Printed English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Memory
Stores up to 100 events and programs
Model
IVD Certified
Noise Level
<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)
Choices of 9 Accel/10 Decel Rates
Program Storage
Up to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor
18 Rotor options available
Rotor Included
No
Run Time
Up to 99 hr. 59 min. 59 sec.
Standards
ISO 14971, ISO 13485, ISO 9001
Temperature Range
-10°C to +40°C, ±2°C
Timer
Option to count up or down and to set “At start” or “At speed”
Type
Benchtop Centrifuge
Ventilated
No
Warranty
5 Year Limited
Dimensions (L x W x H) Exterior
690 x 746 x 361 mm
Length (English) Exterior
27.2 in.
Length (Metric) Exterior
690 mm
Width (English) Exterior
29.4 in.
Width (Metric) Exterior
746 mm
Height (English) Exterior
14.2 in.
Height (English) Exterior Lid Open
33.9 in.
Height (Metric) Exterior
361 mm
Height (Metric) Exterior Lid Open
861 mm
Depth (English)
27.2 in.
Depth (Metric)
690 mm
Net Weight
120 kg
Voltage
120 V
Wattage
1450 w
Frequency
50/60 Hz
Laboratory/Cleanroom Use
Cleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller Type
Microprocessor
Description
Multifuge X4R Pro-MD 220V 60Hz
Capacity
4 x 1000 mL BioBottles, 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 4 x 6 Microplates with TX-1000 Rotor
Max. RCF
25,830 x g Fixed Angle with Microliter 30 x 2 rotor, 7,197 x g Swing-Out with H-Flex HS4 rotor
Product Line
Multifuge
Refrigerated
Yes
Display
Full Color Touch Screen Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
Printed English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the accroding country supply voltage, declaration of conformity
Material
Front, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Memory
Stores up to 100 events and programs
Model
IVD Certified
Noise Level
<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)
Choices of 9 Accel/10 Decel Rates
Program Storage
Up to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor
18 Rotor options available
Rotor Included
No
Run Time
Up to 99 hr. 59 min. 59 sec.
Standards
ISO 14971, ISO 13485, ISO 9001
Temperature Range
-10°C to +40°C, ±2°C
Timer
Option to count up or down and to set “At start” or “At speed”
Type
Benchtop Centrifuge
Ventilated
No
Warranty
5 Year Limited
Dimensions (L x W x H) Exterior
690 x 746 x 361 mm
Length (English) Exterior
27.2 in.
Length (Metric) Exterior
690 mm
Width (English) Exterior
29.4 in.
Width (Metric) Exterior
746 mm
Height (English) Exterior
14.2 in.
Height (English) Exterior Lid Open
33.9 in.
Height (Metric) Exterior
361 mm
Height (Metric) Exterior Lid Open
861 mm
Depth (English)
27.2 in.
Depth (Metric)
690 mm
Net Weight
120 kg
Voltage
220 V
Wattage
1950 w
Frequency
60 Hz
Laboratory/Cleanroom Use
Cleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller Type
Microprocessor
Description
Multifuge X4R Pro 220V 60Hz
Capacity
4 x 1000 mL BioBottles, 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 4 x 6 Microplates with TX-1000 Rotor
Max. RCF
25,830 x g Fixed Angle with Microliter 30 x 2 rotor, 7,197 x g Swing-Out with H-Flex HS4 rotor
Product Line
Multifuge
Refrigerated
Yes
Display
Full Color Touch Screen Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
Printed English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Memory
Stores up to 100 events and programs
Model
General Use Only
Noise Level
<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)
Choices of 9 Accel/10 Decel Rates
Program Storage
Up to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor
18 Rotor options available
Rotor Included
No
Run Time
Up to 99 hr. 59 min. 59 sec.
Standards
ISO 14971, ISO 13485, ISO 9001
Temperature Range
-10°C to +40°C, ±2°C
Timer
Option to count up or down and to set “At start” or “At speed”
Type
Benchtop Centrifuge
Ventilated
No
Warranty
5 Year Limited
Dimensions (L x W x H) Exterior
690 x 746 x 361 mm
Length (English) Exterior
27.2 in.
Length (Metric) Exterior
690 mm
Width (English) Exterior
29.4 in.
Width (Metric) Exterior
746 mm
Height (English) Exterior
14.2 in.
Height (English) Exterior Lid Open
33.9 in.
Height (Metric) Exterior
361 mm
Height (Metric) Exterior Lid Open
861 mm
Depth (English)
27.2 in.
Depth (Metric)
690 mm
Net Weight
120 kg
Voltage
220 V
Wattage
1950 w
Frequency
60 Hz
Laboratory/Cleanroom Use
Cleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller Type
Microprocessor
Description
Multifuge X4 Pro 208-240V 50/60Hz
Capacity
4 x 1000 mL BioBottles, 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 4 x 6 Microplates with TX-1000 Rotor
Max. RCF
25,830 x g Fixed Angle with Microliter 30 x 2 rotor, 7,197 x g Swing-Out with H-Flex HS4 rotor
Product Line
Multifuge
Refrigerated
No
Display
Full Color Touch Screen Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
Printed English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Memory
Stores up to 100 events and programs
Model
General Use Only
Noise Level
<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)
Choices of 9 Accel/10 Decel Rates
Program Storage
Up to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor
18 Rotor options available
Rotor Included
No
Run Time
Up to 99 hr. 59 min. 59 sec.
Standards
ISO 14971, ISO 13485, ISO 9001
Timer
Option to count up or down and to set “At start” or “At speed”
Type
Benchtop Centrifuge
Ventilated
Yes
Warranty
5 Year Limited
Dimensions (L x W x H) Exterior
690 x 566 x 362 mm
Length (English) Exterior
27.2 in.
Length (Metric) Exterior
690 mm
Width (English) Exterior
22.3 in.
Width (Metric) Exterior
566 mm
Height (English) Exterior
14.2 in.
Height (English) Exterior Lid Open
33.9 in.
Height (Metric) Exterior
362 mm
Height (Metric) Exterior Lid Open
861 mm
Depth (English)
27.2 in.
Depth (Metric)
690 mm
Net Weight
89 kg
Voltage
208-240 V
Wattage
1600 w
Frequency
50/60 Hz
Laboratory/Cleanroom Use
Cleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller Type
Microprocessor
Description
Multifuge X4R Pro 220-240V 50Hz/230V 60Hz
Capacity
4 x 1000 mL BioBottles, 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 4 x 6 Microplates with TX-1000 Rotor
Max. RCF
25,830 x g Fixed Angle with Microliter 30 x 2 rotor, 7,197 x g Swing-Out with H-Flex HS4 rotor
Product Line
Multifuge
Refrigerated
Yes
Display
Full Color Touch Screen Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
Printed English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Memory
Stores up to 100 events and programs
Model
General Use Only
Noise Level
<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)
Choices of 9 Accel/10 Decel Rates
Program Storage
Up to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor
18 Rotor options available
Rotor Included
No
Run Time
Up to 99 hr. 59 min. 59 sec.
Standards
ISO 14971, ISO 13485, ISO 9001
Temperature Range
-10°C to +40°C, ±2°C
Timer
Option to count up or down and to set “At start” or “At speed”
Type
Benchtop Centrifuge
Ventilated
No
Warranty
5 Year Limited
Dimensions (L x W x H) Exterior
690 x 746 x 361 mm
Length (English) Exterior
27.2 in.
Length (Metric) Exterior
690 mm
Width (English) Exterior
29.4 in.
Width (Metric) Exterior
746 mm
Height (English) Exterior
14.2 in.
Height (English) Exterior Lid Open
33.9 in.
Height (Metric) Exterior
361 mm
Height (Metric) Exterior Lid Open
861 mm
Depth (English)
27.2 in.
Depth (Metric)
690 mm
Net Weight
120 kg
Voltage
220-240 V
Wattage
1950 w
Frequency
50/60 Hz
Laboratory/Cleanroom Use
Cleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes
Max. RCF
3,374 x g
Max. Speed
3,800 rpm
Product Line
Multifuge
Refrigerated
No
Display
Full Color Touch Screen Interface
Drive System
Direct, Brushless Induction Low Profile
Green Features
Energy efficient, less waste
Includes
Printed English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Memory
Stores up to 100 events and programs
Model
IVD Certified
Profile (Acceleration/Braking)
Choices of 9 Accel/10 Decel Rates
Program Storage
Up to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor
Blood Tube Package Includes TX-1000 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube Adapters
Rotor Included
Yes
Run Time
Up to 99 hr. 59 min. 59 sec.
Standards
ISO 14971, ISO 13485, ISO 9001
Timer
Option to count up or down and to set “At start” or “At speed”
Type
Benchtop Centrifuge
Ventilated
Yes
Warranty
Centrifuge: 5 Year Limited, Rotor: See Manual
Net Weight
See individual components of package
Voltage
120 V
Frequency
60 Hz
Laboratory/Cleanroom Use
Cleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes
Max. RCF
4,122 x g (with TX-1000 rotor)
Max. Speed
4,200 rpm (with TX-1000 rotor)
Product Line
Multifuge
Refrigerated
Yes
Display
Full Color Touch Screen Interface
Drive System
Direct, Brushless Induction Low Profile
Green Features
Energy efficient, less waste
Includes
Printed English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Memory
Stores up to 100 events and programs
Profile (Acceleration/Braking)
Choices of 9 Accel/10 Decel Rates
Program Storage
Up to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor
Blood Tube Package Includes TX-1000 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube Adapters
Rotor Included
Yes
Run Time
Up to 99 hr. 59 min. 59 sec.
Standards
ISO 14971, ISO 13485, ISO 9001
Temperature Range
-10°C to +40°C, ±2°C
Timer
Option to count up or down and to set “At start” or “At speed”
Type
Benchtop Centrifuge
Ventilated
No
Warranty
Centrifuge: 5 Year Limited, Rotor: See Manual
Net Weight
See individual components of package
Voltage
120 V
Frequency
60 Hz
Laboratory/Cleanroom Use
Cleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller Type
Microprocessor
Description
Multifuge X4 Pro-MD 208-240V 50/60Hz
Capacity
4 x 1000 mL BioBottles, 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 4 x 6 Microplates with TX-1000 Rotor
Max. RCF
25,830 x g Fixed Angle with Microliter 30 x 2 Rotor, 7,197 x g Swing-Out with H-Flex HS4 Rotor
Product Line
Multifuge
Refrigerated
No
Display
Full Color Touch Screen Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
Printed English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Memory
Stores up to 100 events and programs
Model
IVD Certified
Noise Level
<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)
Choices of 9 Accel/10 Decel Rates
Program Storage
Up to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor
18 Rotor options available
Rotor Included
No
Run Time
Up to 99 hr. 59 min. 59 sec.
Standards
ISO 14971, ISO 13485, ISO 9001
Timer
Option to count up or down and to set 'At start' or 'At speed'
Type
Benchtop Centrifuge
Ventilated
Yes
Warranty
5 Year Limited
Dimensions (L x W x H) Exterior
690 x 566 x 362 mm
Length (English) Exterior
27.2 in.
Length (Metric) Exterior
690 mm
Width (English) Exterior
22.3 in.
Width (Metric) Exterior
566 mm
Height (English) Exterior
14.2 in.
Height (English) Exterior Lid Open
33.9 in.
Height (Metric) Exterior
362 mm
Height (Metric) Exterior Lid Open
861 mm
Depth (English)
27.2 in.
Depth (Metric)
690 mm
Net Weight
89 kg
Voltage
208/240 V
Wattage
1600 w
Frequency
50/60 Hz
Laboratory/Cleanroom Use
Cleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller Type
Microprocessor
Description
Multifuge X4 Pro-MD 120V
Capacity
4 x 1000 mL BioBottles, 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 4 x 6 Microplates with TX-1000 Rotor
Max. RCF
25,830 x g Fixed Angle with Microliter 30 x 2 Rotor, 7,197 x g Swing-Out with H-Flex HS4 Rotor
Product Line
Multifuge
Refrigerated
No
Display
Full Color Touch Screen Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
Printed English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Memory
Stores up to 100 events and programs
Model
IVD Certified
Noise Level
<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)
Choices of 9 Accel/10 Decel Rates
Program Storage
Up to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor
18 Rotor options available
Rotor Included
No
Run Time
Up to 99 hr. 59 min. 59 sec.
Standards
ISO 14971, ISO 13485, ISO 9001
Timer
Option to count up or down and to set 'At start' or 'At speed'
Type
Benchtop Centrifuge
Ventilated
Yes
Warranty
5 Year Limited
Dimensions (L x W x H) Exterior
690 x 566 x 362 mm
Length (English) Exterior
27.2 in.
Length (Metric) Exterior
690 mm
Width (English) Exterior
22.3 in.
Width (Metric) Exterior
566 mm
Height (English) Exterior
14.2 in.
Height (English) Exterior Lid Open
33.9 in.
Height (Metric) Exterior
362 mm
Height (Metric) Exterior Lid Open
861 mm
Depth (English)
27.2 in.
Depth (Metric)
690 mm
Net Weight
89 kg
Voltage
120 V
Wattage
1300 w
Frequency
50/60 Hz
Laboratory/Cleanroom Use
Cleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller Type
Microprocessor
Description
Multifuge X4R Pro-MD 220-240V 50Hz / 230V 60Hz
Capacity
4 x 1000 mL BioBottles, 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 4 x 6 Microplates with TX-1000 Rotor
Max. RCF
25,830 x g Fixed Angle with Microliter 30 x 2 rotor, 7,197 x g Swing-Out with H-Flex HS4 rotor
Product Line
Multifuge
Refrigerated
Yes
Display
Full Color Touch Screen Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
Printed English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Memory
Stores up to 100 events and programs
Model
IVD Certified
Noise Level
<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)
Choices of 9 Accel/10 Decel Rates
Program Storage
Up to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor
18 Rotor options available
Rotor Included
No
Run Time
Up to 99 hr. 59 min. 59 sec.
Standards
ISO 14971, ISO 13485, ISO 9001
Temperature Range
-10°C to +40°C, ±2°C
Timer
Option to count up or down and to set “At start” or “At speed”
Type
Benchtop Centrifuge
Ventilated
No
Warranty
5 Year Limited
Dimensions (L x W x H) Exterior
690 x 746 x 361 mm
Length (English) Exterior
27.2 in.
Length (Metric) Exterior
690 mm
Width (English) Exterior
29.4 in.
Width (Metric) Exterior
746 mm
Height (English) Exterior
14.2 in.
Height (English) Exterior Lid Open
33.9 in.
Height (Metric) Exterior
361 mm
Height (Metric) Exterior Lid Open
861 mm
Depth (English)
27.2 in.
Depth (Metric)
690 mm
Net Weight
120 kg
Voltage
220-240 V
Wattage
1950 w
Frequency
50/60 Hz
Laboratory/Cleanroom Use
Cleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller Type
Microprocessor
Description
Multifuge X4R Pro-MD 120V
Capacity
4 x 1000 mL BioBottles, 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 4 x 6 Microplates with TX-1000 Rotor
Max. RCF
25,830 x g Fixed Angle with Microliter 30 x 2 rotor, 7,197 x g Swing-Out with H-Flex HS4 rotor
Product Line
Multifuge
Refrigerated
Yes
Display
Full Color Touch Screen Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
Printed English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Memory
Stores up to 100 events and programs
Model
IVD Certified
Noise Level
<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)
Choices of 9 Accel/10 Decel Rates
Program Storage
Up to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor
18 Rotor options available
Rotor Included
No
Run Time
Up to 99 hr. 59 min. 59 sec.
Standards
ISO 14971, ISO 13485, ISO 9001
Temperature Range
-10°C to +40°C, ±2°C
Timer
Option to count up or down and to set “At start” or “At speed”
Type
Benchtop Centrifuge
Ventilated
No
Warranty
5 Year Limited
Dimensions (L x W x H) Exterior
690 x 746 x 361 mm
Length (English) Exterior
27.2 in.
Length (Metric) Exterior
690 mm
Width (English) Exterior
29.4 in.
Width (Metric) Exterior
746 mm
Height (English) Exterior
14.2 in.
Height (English) Exterior Lid Open
33.9 in.
Height (Metric) Exterior
361 mm
Height (Metric) Exterior Lid Open
861 mm
Depth (English)
27.2 in.
Depth (Metric)
690 mm
Net Weight
120 kg
Voltage
120 V
Wattage
1450 w
Frequency
50/60 Hz
Laboratory/Cleanroom Use
Cleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller Type
Microprocessor
Description
Multifuge X4R Pro-MD 220V 60Hz
Capacity
4 x 1000 mL BioBottles, 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 4 x 6 Microplates with TX-1000 Rotor
Max. RCF
25,830 x g Fixed Angle with Microliter 30 x 2 rotor, 7,197 x g Swing-Out with H-Flex HS4 rotor
Product Line
Multifuge
Refrigerated
Yes
Display
Full Color Touch Screen Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
Printed English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the accroding country supply voltage, declaration of conformity
Material
Front, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Memory
Stores up to 100 events and programs
Model
IVD Certified
Noise Level
<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)
Choices of 9 Accel/10 Decel Rates
Program Storage
Up to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor
18 Rotor options available
Rotor Included
No
Run Time
Up to 99 hr. 59 min. 59 sec.
Standards
ISO 14971, ISO 13485, ISO 9001
Temperature Range
-10°C to +40°C, ±2°C
Timer
Option to count up or down and to set “At start” or “At speed”
Type
Benchtop Centrifuge
Ventilated
No
Warranty
5 Year Limited
Dimensions (L x W x H) Exterior
690 x 746 x 361 mm
Length (English) Exterior
27.2 in.
Length (Metric) Exterior
690 mm
Width (English) Exterior
29.4 in.
Width (Metric) Exterior
746 mm
Height (English) Exterior
14.2 in.
Height (English) Exterior Lid Open
33.9 in.
Height (Metric) Exterior
361 mm
Height (Metric) Exterior Lid Open
861 mm
Depth (English)
27.2 in.
Depth (Metric)
690 mm
Net Weight
120 kg
Voltage
220 V
Wattage
1950 w
Frequency
60 Hz
Laboratory/Cleanroom Use
Cleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller Type
Microprocessor
Description
Multifuge X4R Pro 220V 60Hz
Capacity
4 x 1000 mL BioBottles, 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 4 x 6 Microplates with TX-1000 Rotor
Max. RCF
25,830 x g Fixed Angle with Microliter 30 x 2 rotor, 7,197 x g Swing-Out with H-Flex HS4 rotor
Product Line
Multifuge
Refrigerated
Yes
Display
Full Color Touch Screen Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
Printed English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Memory
Stores up to 100 events and programs
Model
General Use Only
Noise Level
<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)
Choices of 9 Accel/10 Decel Rates
Program Storage
Up to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor
18 Rotor options available
Rotor Included
No
Run Time
Up to 99 hr. 59 min. 59 sec.
Standards
ISO 14971, ISO 13485, ISO 9001
Temperature Range
-10°C to +40°C, ±2°C
Timer
Option to count up or down and to set “At start” or “At speed”
Type
Benchtop Centrifuge
Ventilated
No
Warranty
5 Year Limited
Dimensions (L x W x H) Exterior
690 x 746 x 361 mm
Length (English) Exterior
27.2 in.
Length (Metric) Exterior
690 mm
Width (English) Exterior
29.4 in.
Width (Metric) Exterior
746 mm
Height (English) Exterior
14.2 in.
Height (English) Exterior Lid Open
33.9 in.
Height (Metric) Exterior
361 mm
Height (Metric) Exterior Lid Open
861 mm
Depth (English)
27.2 in.
Depth (Metric)
690 mm
Net Weight
120 kg
Voltage
220 V
Wattage
1950 w
Frequency
60 Hz
Laboratory/Cleanroom Use
Cleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller Type
Microprocessor
Description
Multifuge X4 Pro 208-240V 50/60Hz
Capacity
4 x 1000 mL BioBottles, 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 4 x 6 Microplates with TX-1000 Rotor
Max. RCF
25,830 x g Fixed Angle with Microliter 30 x 2 rotor, 7,197 x g Swing-Out with H-Flex HS4 rotor
Product Line
Multifuge
Refrigerated
No
Display
Full Color Touch Screen Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
Printed English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Memory
Stores up to 100 events and programs
Model
General Use Only
Noise Level
<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)
Choices of 9 Accel/10 Decel Rates
Program Storage
Up to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor
18 Rotor options available
Rotor Included
No
Run Time
Up to 99 hr. 59 min. 59 sec.
Standards
ISO 14971, ISO 13485, ISO 9001
Timer
Option to count up or down and to set “At start” or “At speed”
Type
Benchtop Centrifuge
Ventilated
Yes
Warranty
5 Year Limited
Dimensions (L x W x H) Exterior
690 x 566 x 362 mm
Length (English) Exterior
27.2 in.
Length (Metric) Exterior
690 mm
Width (English) Exterior
22.3 in.
Width (Metric) Exterior
566 mm
Height (English) Exterior
14.2 in.
Height (English) Exterior Lid Open
33.9 in.
Height (Metric) Exterior
362 mm
Height (Metric) Exterior Lid Open
861 mm
Depth (English)
27.2 in.
Depth (Metric)
690 mm
Net Weight
89 kg
Voltage
208-240 V
Wattage
1600 w
Frequency
50/60 Hz
Laboratory/Cleanroom Use
Cleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
EN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
Controller Type
Microprocessor
Description
Multifuge X4R Pro 220-240V 50Hz/230V 60Hz
Capacity
4 x 1000 mL BioBottles, 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 4 x 6 Microplates with TX-1000 Rotor
Max. RCF
25,830 x g Fixed Angle with Microliter 30 x 2 rotor, 7,197 x g Swing-Out with H-Flex HS4 rotor
Product Line
Multifuge
Refrigerated
Yes
Display
Full Color Touch Screen Interface
Drive System
Direct, Brushless Induction Low Profile
For Use With (Equipment)
Purchase of Thermo Scientific rotor and accessories required for operation
Green Features
Energy efficient, less waste
Includes
Printed English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Memory
Stores up to 100 events and programs
Model
General Use Only
Noise Level
<64 dB(A) Rotor F15-8x50c; 14500 RPM
Profile (Acceleration/Braking)
Choices of 9 Accel/10 Decel Rates
Program Storage
Up to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor
18 Rotor options available
Rotor Included
No
Run Time
Up to 99 hr. 59 min. 59 sec.
Standards
ISO 14971, ISO 13485, ISO 9001
Temperature Range
-10°C to +40°C, ±2°C
Timer
Option to count up or down and to set “At start” or “At speed”
Type
Benchtop Centrifuge
Ventilated
No
Warranty
5 Year Limited
Dimensions (L x W x H) Exterior
690 x 746 x 361 mm
Length (English) Exterior
27.2 in.
Length (Metric) Exterior
690 mm
Width (English) Exterior
29.4 in.
Width (Metric) Exterior
746 mm
Height (English) Exterior
14.2 in.
Height (English) Exterior Lid Open
33.9 in.
Height (Metric) Exterior
361 mm
Height (Metric) Exterior Lid Open
861 mm
Depth (English)
27.2 in.
Depth (Metric)
690 mm
Net Weight
120 kg
Voltage
220-240 V
Wattage
1950 w
Frequency
50/60 Hz
Laboratory/Cleanroom Use
Cleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes
Max. RCF
3,374 x g
Max. Speed
3,800 rpm
Product Line
Multifuge
Refrigerated
No
Display
Full Color Touch Screen Interface
Drive System
Direct, Brushless Induction Low Profile
Green Features
Energy efficient, less waste
Includes
Printed English user manual, USB stick with 28 languages of user manual, corrosion resistance oil (70009824), power cord for the according country supply voltage, declaration of conformity
Material
Front, top, sides: PC-ABS plastic painted with 2K PUR-based lacquer; metal housing: cold rolled steel with powder paint; centrifuge bowl: stainless steel; centrifuge feet: rubber
Memory
Stores up to 100 events and programs
Model
IVD Certified
Profile (Acceleration/Braking)
Choices of 9 Accel/10 Decel Rates
Program Storage
Up to 100 Programs, including multi-step programming via Touch Screen; Including programmabile Pulse options
Rotor
Blood Tube Package Includes TX-1000 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube Adapters
Rotor Included
Yes
Run Time
Up to 99 hr. 59 min. 59 sec.
Standards
ISO 14971, ISO 13485, ISO 9001
Timer
Option to count up or down and to set “At start” or “At speed”
Type
Benchtop Centrifuge
Ventilated
Yes
Warranty
Centrifuge: 5 Year Limited, Rotor: See Manual
Net Weight
See individual components of package
Voltage
120 V
Frequency
60 Hz
Laboratory/Cleanroom Use
Cleanroom compatibility Class ISO 7 according to DIN EN ISO 14644-1
Unit Size
Each
Showing 10 of 18Show more
The GP Pro centrifuge series is available as ventilated models (X4) or refrigerated models (X4R) as well as for general laboratory use or IVD certified.
Performance features:
Intuitive touch screen user interface simplifies operation with easily accessible menu bar and centrifuge health icon for at-a-glance monitoring
Thermo Scientific™ Accumulated Centrifugal Effect (ACE™) integrator function for increased reproducibility
Thermo Scientific™ SMARTSpin™ imbalance technology optimizes safety and maximizes acceleration, braking and residual load imbalance for each rotor and bucket option
Thermo Scientific™ Auto-Lock™ Rotor Exchange:
Secure, ergonomic push-button rotor exchange in as little as three seconds with no tools required
Full chamber access for cleaning convenience and minimizes risk of contamination
Flexibility to quickly switch between applications and evolve changing laboratory needs
Biocontainment sealing options for glove-friendly, one-handed open/close capability are ergonomically designed for both right and left-handed operations
Audible and visual closure cues enhance safety and security
Biocontainment certification by National Institute for Health Protection, Porton Down, UK or TÜV NORD CERT GmbH, Germany
Rotor versatility and Thermo Scientific™ Fiberlite™ Carbon Fiber Rotors:
Up to 18 unique rotor options, including Fiberlite Carbon Fiber Rotor options, to help maximize performance and flexibility
Fiberlite carbon fiber rotors are supported by a 15-year warranty and no limitation in cycles of use
Carbon fiber offers a unique ability to mold each cavity to perfectly fit disposable conical tubes, bottles, and other consumables
Safety and ergonomic features:
Space saving design with compact dimensions to optimize work area at an ergonomic height
Lid safety features promote ergonomic sample loading and unloading and enable easy cleaning
Energy efficiency:
System offers up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tube processing
Scheduling and compressor on/off capability promotes energy savings
Fiberlite rotors weigh up to 60% less than metal equivalent rotors and therefore accelerate/decelerate faster translating into reduced energy consumption per run*
Connectivity and data management:
Event Log, Rotor Log and Chart Data can be exported via USB in CSV or PDF formats
Programs are downloadable and shareable between units
Front USB port for easier plug in and data transfer
Optional Thermo Scientific™ SECURE-Spin™ Mode available, enables advanced and secure control features in the X Pro centrifuges by supporting 21 CFR Part 11 data management compliance and traceability
Traceable documentation provides audit trail for setting and program changes
Compatible with Thermo Scientific™ Centri-Log™ Plus Data Management Software enabling remote monitoring of unlimited number of centrifuges and sample tracking with a barcode reader
Previous models
Multifuge X3, Multifuge X3R
*Compare data highlighted in the Green Fact Sheet from 2021 under https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/PG2030-PJT4622-COL33722-Centrifuges-FactSheet-Americas-FHR.pdf