General Purpose Centrifuge Rotor Packages
Actual product may vary
Thermo Scientific™
General Purpose Centrifuge Rotor Packages
Everything you need for your swing out rotor in one package
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionTypeCapacity
75016111Thermo Scientific TX-200 Blood Tube Package with TX-200 Swinging Bucket RotorTX-200 with 4 Buckets, 4 ClickSeal Biocontainment lids, 5/7 and 10 mL Blood Tube AdaptersMax in TX-200: 36 x 5/7 ml Blood Tubes, 28 x 10 mL Blood tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors)
75016103Thermo Scientific TX-1000 Blood Tube PackageTX-1000 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube AdaptersMax in TX-1000: 196 x 5/7 mL Blood Tubes, 148 x 9/10 mL Blood Tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors)
75016104Thermo Scientific TX-1000 Microplate PackageTX-1000 with Buckets, ClickSeal Biocontainment lids, microplate AdaptersMax in TX-1000: 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 Rotors)
75016105Thermo Scientific TX-1000 Cell Culture PackageTX-1000 with Buckets, ClickSeal Biocontainment lids, 15 and 50 mL conical tube adaptersMax in TX-1000: 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors)
75016106Thermo Scientific TX-200 Cell Culture PackageTX-200 with Buckets, ClickSeal Biocontainment lids, 15 and 50 mL conical tube adaptersMax in TX-200: 20 x 15 mL Conical Tubes, 4 x 50 mL Conical Tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors)
75016107Thermo Scientific TX-400 Blood Tube PackageTX-400 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube AdaptersMax in TX-400: 76 x 5/7 mL Blood Tubes, 56 x 9/10 mL Blood Tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors)
75016108Thermo Scientific TX-400 Cell Culture PackageTX-400 with Buckets, ClickSeal Biocontainment lids, 15 and 50 mL conical tube adaptersMax in TX-400: 36 x 15 mL Conical Tubes, 16 x 50 mL Conical Tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors)
75016109Thermo Scientific TX-750 Blood Tube PackageTX-750 with Buckets, ClickSeal Biocontainment lids, 5/7 and 9/10 mL Blood Tube AdaptersMax in TX-750: 148 x 5/7 mL Blood Tubes, 84 x 9/10 mL Blood Tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors)
75016110Thermo Scientific TX-750 Cell Culture PackageTX-750 with Buckets, ClickSeal Biocontainment lids, 15 and 50 mL conical tube adaptersMax in TX-750: 56 x 15 mL Conical Tubes, 28 x 50 mL Conical Tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors)
9 Options
Catalog number 75016111
Price (USD)/ Each
-
Request A Quote
Description:
Thermo Scientific TX-200 Blood Tube Package with TX-200 Swinging Bucket Rotor
Type:
TX-200 with 4 Buckets, 4 ClickSeal Biocontainment lids, 5/7 and 10 mL Blood Tube Adapters
Capacity:
Max in TX-200: 36 x 5/7 ml Blood Tubes, 28 x 10 mL Blood tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors)
General Purpose Centrifuge Rotor packages with all the pieces you need to start spinning right away, including a rotor cross, bucket, lids and adapters
Specifications
DescriptionThermo Scientific TX-200 Blood Tube Package with TX-200 Swinging Bucket Rotor
MaterialSee individual centrifuge, rotor and accessory data
TypeTX-200 with 4 Buckets, 4 ClickSeal Biocontainment lids, 5/7 and 10 mL Blood Tube Adapters
CapacityMax in TX-200: 36 x 5/7 ml Blood Tubes, 28 x 10 mL Blood tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors)
Net WeightSee individual components of package
For Use With (Equipment)Purchase of Thermo Scientific General Purpose Centrifuge required for operation
Certifications/ComplianceEN/UL/IEC 61010-1EN/UL/IEC 61010-2EN/UL/IEC 61326-1EN/UL/IEC 61326-2FCC part 15
IncludesPackage Includes: TX-200 rotor cross w/ Auto=Lock, 4 Buckets, 4 ClickSeal biocontainment lids, 5/7mL blood tube adapters (set/4, 36 tube capacity), 10mL blood tube adapters (set/4, 28 tube capacity)
WarrantySee Manual
BiocontainmentYes
Unit SizeEach