Sorvall ST1 R Plus-MD, 208-240V H-Flex Blood Tube Package
Sorvall ST1 R Plus-MD, 208-240V H-Flex Blood Tube Package
Thermo Scientific™
Sorvall ST1 R Plus-MD, 208-240V H-Flex Blood Tube Package
Green features
Waste no time finding the right part numbers for your application! The application driven centrifuge package ensures convenient, fast ordering of all components necessary to run a specific protocol.
Have Questions?
Catalog number 75017021
Price (USD)/ Each
-
Request A Quote
Includes:
Part Numbers In Pk. 75009380 (centrifuge),75003300(rotor),75003301(set of buckets),75003302(lids),75003307(set of adapters),75003305(set of adapters)
Waste no time finding the right part numbers for your application! The application driven centrifuge package ensures convenient, fast ordering of all components necessary to run a specific protocol. Run microplates for PCR and blood tubes for antibody testing used in COVID-19 diagnosis protocols, all in the same rotor without changing rotors or buckets. Contains centrifuge, H-Flex rotor, buckets, biocontainment lids, 5/7 mL blood tube adapter (maximum capacity 84 x 5/7 mL), 10 mL blood tube adapters (maximum capacity 66 x 10 mL) , microplate adapters (maximum capacity 10 standard, 2 deepwell).
  • Easy to Use:
    • Intuitive controls simplify operation and provide detailed information on a wide range of operating parameters and processing status for optimal performance.
    • Modern industrial design supports continuous improvement in rotor loading ergonomics, user safety and current needed certifications
    • Motorized lid latch enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches.
    Thermo Scientific™ Auto-Lock™ III Rotor System:
    • Secure locking system allows easy push-button installation and exchange of rotors.
    • System saves time when switching between applications.
    • Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life.
    Thermo Scientific™ ClickSeal™ Bucket Sealing System:
    • Biocontainment solution seals with a snap; eliminates screw caps and complicated clips.
    • System has glove-friendly, one-handed open/close capability.
    • System is certified by CAMR™ in Porton Down, UK.
    Thermo Scientific™ SMARTSpin™ Technology:
    • Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option:
    • Technology optimizes safety and improves separations.
    Space-Saving Design:
    • Compact dimensions optimize work area and ergonomic height and curved design ensure easy sample loading and unloading and easy cleaning.
    Energy Efficient:
    • System offers up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing.
Specifications
Drive SystemDirect, Brushless induction low profile motor with choices of 9 acceleration, 10 deceleration rates
Green FeaturesLess waste, Energy efficient
IncludesPart Numbers In Pk. 75009380 (centrifuge),75003300(rotor),75003301(set of buckets),75003302(lids),75003307(set of adapters),75003305(set of adapters)
TypeGeneral Purpose Centrifuge Series
Unit SizeEach