Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Additive | Contains trace amount of surfactant to inhibit agglomeration and promote stability |
Concentration | 10% |
Description | Particles useful as model systems for fluid mechanics experiments and as challenge particles for large pore filtration systems. |
Color | White or Colorless |
For Use With (Application) | Fluid Mechanics, Large Pore Filtration |
Material | Borosilicate Glass |
Product Type | Copolymer Microsphere Suspensions |
Includes | General product handling insert sheet |
Packaging | 1000 mL |
Particle Density | 1.05g/cm3 |
Quantity | 1000 mL |
Refractive Index | 1.59 at 589nm (25°C) |
Stability | Product stable for a minimum of 24 months |
Diameter (Metric) | 7.9 μm |
Unit Size | Each |
Additive | Contains trace amount of surfactant to inhibit agglomeration and promote stability |
Concentration | 10% |
Description | Particles useful as model systems for fluid mechanics experiments and as challenge particles for large pore filtration systems. |
Color | White or Colorless |
For Use With (Application) | Fluid Mechanics, Large Pore Filtration |
Material | Polystyrene Divinylbenzene (PS-DVB) |
Product Type | Copolymer Microsphere Suspensions |
Includes | General product handling insert sheet |
Packaging | 15 mL |
Particle Density | 1.05g/cm3 |
Quantity | 15 mL |
Refractive Index | 1.59 at 589nm (25°C) |
Stability | Product stable for a minimum of 24 months |
Diameter (Metric) | 3.2 μm |
Unit Size | Each |
Additive | Contains trace amount of surfactant to inhibit agglomeration and promote stability |
Concentration | 10% |
Description | Particles useful as model systems for fluid mechanics experiments and as challenge particles for large pore filtration systems. |
Color | White or Colorless |
For Use With (Application) | Fluid Mechanics, Large Pore Filtration |
Material | Polystyrene Divinylbenzene (PS-DVB) |
Product Type | Copolymer Microsphere Suspensions |
Includes | General product handling insert sheet |
Packaging | 100 mL |
Particle Density | 1.05g/cm3 |
Quantity | 100 mL |
Refractive Index | 1.59 at 589nm (25°C) |
Stability | Product stable for a minimum of 24 months |
Diameter (Metric) | 3.2 μm |
Unit Size | Each |
Additive | Contains trace amount of surfactant to inhibit agglomeration and promote stability |
Concentration | 10% |
Description | Particles useful as model systems for fluid mechanics experiments and as challenge particles for large pore filtration systems. |
Color | White or Colorless |
For Use With (Application) | Fluid Mechanics, Large Pore Filtration |
Material | Polystyrene Divinylbenzene (PS-DVB) |
Product Type | Copolymer Microsphere Suspensions |
Includes | General product handling insert sheet |
Packaging | 1000 mL |
Particle Density | 1.05g/cm3 |
Quantity | 1000 mL |
Refractive Index | 1.59 at 589nm (25°C) |
Stability | Product stable for a minimum of 24 months |
Diameter (Metric) | 3.2 μm |
Unit Size | Each |
Additive | Contains trace amount of surfactant to inhibit agglomeration and promote stability |
Concentration | 10% |
Description | Particles useful as model systems for fluid mechanics experiments and as challenge particles for large pore filtration systems. |
Color | White or Colorless |
For Use With (Application) | Fluid Mechanics, Large Pore Filtration |
Material | Polystyrene Divinylbenzene (PS-DVB) |
Product Type | Copolymer Microsphere Suspensions |
Includes | General product handling insert sheet |
Packaging | 15 mL |
Particle Density | 1.05g/cm3 |
Quantity | 15 mL |
Refractive Index | 1.59 at 589nm (25°C) |
Stability | Product stable for a minimum of 24 months |
Diameter (Metric) | 6 μm |
Unit Size | Each |
Additive | Contains trace amount of surfactant to inhibit agglomeration and promote stability |
Concentration | 10% |
Description | Particles useful as model systems for fluid mechanics experiments and as challenge particles for large pore filtration systems. |
Color | White or Colorless |
For Use With (Application) | Fluid Mechanics, Large Pore Filtration |
Material | Polystyrene Divinylbenzene (PS-DVB) |
Product Type | Copolymer Microsphere Suspensions |
Includes | General product handling insert sheet |
Packaging | 100 mL |
Particle Density | 1.05g/cm3 |
Quantity | 100 mL |
Refractive Index | 1.59 at 589nm (25°C) |
Stability | Product stable for a minimum of 24 months |
Diameter (Metric) | 6 μm |
Unit Size | Each |
Additive | Contains trace amount of surfactant to inhibit agglomeration and promote stability |
Concentration | 10% |
Description | Particles useful as model systems for fluid mechanics experiments and as challenge particles for large pore filtration systems. |
Color | White or Colorless |
For Use With (Application) | Fluid Mechanics, Large Pore Filtration |
Material | Polystyrene Divinylbenzene (PS-DVB) |
Product Type | Copolymer Microsphere Suspensions |
Includes | General product handling insert sheet |
Packaging | 1000 mL |
Particle Density | 1.05g/cm3 |
Quantity | 1000 mL |
Refractive Index | 1.59 at 589nm (25°C) |
Stability | Product stable for a minimum of 24 months |
Diameter (Metric) | 6 μm |
Unit Size | Each |
Additive | Contains trace amount of surfactant to inhibit agglomeration and promote stability |
Concentration | 10% |
Description | Particles useful as model systems for fluid mechanics experiments and as challenge particles for large pore filtration systems. |
Color | White or Colorless |
For Use With (Application) | Fluid Mechanics, Large Pore Filtration |
Material | Polystyrene Divinylbenzene (PS-DVB) |
Product Type | Copolymer Microsphere Suspensions |
Includes | General product handling insert sheet |
Packaging | 15 mL |
Particle Density | 1.05g/cm3 |
Quantity | 15 mL |
Refractive Index | 1.59 at 589nm (25°C) |
Stability | Product stable for a minimum of 24 months |
Diameter (Metric) | 7.9 μm |
Unit Size | Each |
Additive | Contains trace amount of surfactant to inhibit agglomeration and promote stability |
Certifications/Compliance | ISO 24450, DIN 12353 |
Concentration | 10% |
Description | Particles useful as model systems for fluid mechanics experiments and as challenge particles for large pore filtration systems. |
Color | White or Colorless |
For Use With (Application) | Fluid Mechanics, Large Pore Filtration |
Material | Borosilicate Glass |
Product Type | Copolymer Microsphere Suspensions |
Includes | General product handling insert sheet |
Packaging | 100 mL |
Particle Density | 1.05g/cm3 |
Quantity | 100 mL |
Refractive Index | 1.59 at 589nm (25°C) |
Stability | Product stable for a minimum of 24 months |
Diameter (Metric) | 7.9 μm |
Unit Size | Each |
Additive | Contains trace amount of surfactant to inhibit agglomeration and promote stability |
Concentration | 10% |
Description | Particles useful as model systems for fluid mechanics experiments and as challenge particles for large pore filtration systems. |
Color | White or Colorless |
For Use With (Application) | Fluid Mechanics, Large Pore Filtration |
Material | Polystyrene Divinylbenzene (PS-DVB) |
Product Type | Copolymer Microsphere Suspensions |
Includes | General product handling insert sheet |
Packaging | 15 mL |
Particle Density | 1.05g/cm3 |
Quantity | 15 mL |
Refractive Index | 1.59 at 589nm (25°C) |
Stability | Product stable for a minimum of 24 months |
Diameter (Metric) | 12 μm |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Diameter (Metric) | Quantity | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
7508C | Each | 7.9 μm | 1000 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
7503A | Each | 3.2 μm | 15 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
7503B | Each | 3.2 μm | 100 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
7503C | Each | 3.2 μm | 1000 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
7505A | Each | 6 μm | 15 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
7505B | Each | 6 μm | 100 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
7505C | Each | 6 μm | 1000 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
7508A | Each | 7.9 μm | 15 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
7508B | Each | 7.9 μm | 100 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
7510A | Each | 12 μm | 15 mL | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Suspensions of large copolymer microspheres are composed of polystyrene polymer, cross-linked with 4 to 8% divinylbenzene (DVB). The particles are chemically inert and can be washed with alcohol and vacuum or air dried. The polymer density is 1.05g/cm3 and the index of refraction is 1.59 at 589nm. They are packaged as aqueous suspensions at 10% solids by weight.
Ordering Information:
15mL, 100mL and 1000mL bottle sizes available. 15mL (“A” Part No. bottles), 100mL (“B” bottles) and 1000mL (“C” bottles) are packaged to order; example (Part No. 7503B and 7503C). Please allow 10 working days when ordering these fill volumes.
Includes:
Material Safety Data Sheet (MSDS) and general product handling insert sheet
Recommended for:
High concentration polymer particles > 3μm used in filter evaluation and testing, fluid mechanics research, dispersion studies and many other research and development projects. Not intended for use in instrument calibration or diagnostic reagents because they lack the exacting specifications needed for those applications. Also useful as experimental particles for acoustical and optical analytical systems.
Not intended for use in instrument calibration or diagnostic reagents.