Thermo Scientific™

Mammalian beta-Galactosidase Assay Kit

Catalog number: 75707
Have Questions?
Thermo Scientific™

Mammalian beta-Galactosidase Assay Kit

Catalog number: 75707
Have Questions?
Catalog Number
75707
Unit Size
75 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
7570775 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific Mammalian β-Galactosidase Assay Kit provides a simple colorimetric method for lysing cultured mammalian cells and measuring beta-Gal activity.

Features of the Mammalian β-Galactosidase Assay Kit:

• Choose stand-alone assay reagent or complete kit for cell lysis and detection
• Colorimetric assay easily measured in a plate reader or spectrophotometer (405nm)
• Adaptable to 96-well microplates or tube assay formats
• Perform homogeneous screening assays or measure activity secondarily after cell lysis
• Also compatiible with prepared lysates from other species such as bacteria

The Assay Reagent and Kit comprise a simple and versatile assay system for measuring β-galactosidase activity. Mammalian cells grown on a plate may be lysed and tested for β-galactosidase activity by adding a single, all-in-one lysis-and-assay solution. Alternatively, the included Thermo Scientific M-PER Mammalian Protein Extraction Reagent may be used to prepare lysate from washed adherent or pelleted cells, and then a portion of the lysate used to quantitate the β-Gal activity. With M-PER Reagent, there is no need to scrape cells or to perform freeze-thaw cycles. The remaining lysates prepared with M-PER Reagent may be used for applications such as reporter assays (luciferase, β-galactosidase and chloramphenical acetyltransferase), protein kinase assays (PKA, PKC, and Tyrosine Kinase), immunoassays (Western blot, ELISA, RIA) and protein purification.

Related Products
beta-Galactosidase Assay Reagent
beta-Galactosidase Assay Stop Solution
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Description
Pierce Mammalian beta-Galactosidase Assay Kit
Detection Method
Reporter Enzyme
For Use With (Application)
Absorbance
For Use With (Equipment)
Microplate Reader
Product Type
Beta-Galactosidase Assay Kit
Dye Type
Enzyme Labeled
Format
96-well Plate
Product Line
Pierce™
Quantity
75 mL Kit
Substrate
Proprietary
Substrate Type
Beta-Gal Substrate
Sufficient For
250 microplate wells of adherent cells
Target
Beta-Galactosidase

Contents & Storage

Sufficient For: 250 microplate wells of adherent cells
  • Pierce beta-Galactosidase Assay Reagent, 25 mL
  • M-PER Mammalian Protein Extraction Reagent, 25 mL
  • Stop Solution, 25 mL

    Store the β-Galactosidase Assay Reagent at -20°C. Store all other components at room temperature.
  • Figures

    Documents & Downloads

    Certificates

      Frequently asked questions (FAQs)

      Citations & References

      Search citations by name, author, journal title or abstract text