BOVIGAM™ Tuberculin PPD Stimulating Antigen, avian (25,000 I.U./mL)
Regulatory requirements vary by country; products may not be available in your geographic area.
Product Image
Thermo Scientific™
BOVIGAM™ Tuberculin PPD Stimulating Antigen, avian (25,000 I.U./mL)
Avian Tuberculin PPD is used as a stimulation control for the BOVIGAM® test. Avian PPD is added to the bloodRead more
Have Questions?
Catalog NumberQuantity
76000655 mL
Catalog number 7600065
Price (USD)
-
Quantity:
5 mL
Ask our AI about this Product
Avian Tuberculin PPD is used as a stimulation control for the BOVIGAM® test. Avian PPD is added to the blood sample to control for unspecific stimulation of blood lymphocytes.
Specifications
Product LineBOVIGAM™, Prionics™
Target SpeciesBovine
Quantity5 mL
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Los clientes que vieron este artículo también vieron
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
2500791BGBCertificate of AnalysisJun 14, 20257600065
2500791BGACertificate of AnalysisMay 12, 20257600065
2500791BGCertificate of AnalysisMar 17, 20257600065
2404696BGACertificate of AnalysisJan 28, 20257600065
2404696BGCertificate of AnalysisNov 14, 20247600065
5 results displayed, search above for a specific certificate
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection