Replacement HEPA Main Filter
Replacement HEPA Main Filter
Replacement HEPA Main Filter
Replacement HEPA Main Filter
Replacement HEPA Main Filter
Replacement HEPA Main Filter
Replacement HEPA Main Filter
Thermo Scientific™
Replacement HEPA Main Filter
For Use With: For Water Jacket models, Steri-cycle and direct heat models (31X, 32X, 37X & 38X) and 8000 Series Incubators
Have Questions?
Catalog NumberType
760175HEPA Filter
Catalog number 760175
Price (USD)
97.18
Online Exclusive
113.00
Save 15.82 (14%)
Each
In stock
Add to cart
Type:
HEPA Filter
Price (USD)
97.18
Online Exclusive
113.00
Save 15.82 (14%)
Each
Add to cart
Help ensure measurable Class 100 air quality performance with Thermo Scientific™ Replacement HEPA Filters and Cleaning Kits.
HEPA Filters
  • 99.97% filtering at 0.3μm
  • Compatible with all Thermo Scientific™ Forma™ Series II 3110, Thermo Scientific™ Steri-Cycle™, and Forma™ 310 Series (with HEPA filtration system option) incubators
  • Main chamber filter available as a stand-alone or packaged in a group with the other chamber in-line filters
Specifications
DescriptionReplacement HEPA Filter
For Use With (Equipment)Forma™ Steri-Cycle™, Direct Heat and Water Jacket
LightingLED
TypeHEPA Filter
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection