Informational:Regulatory requirements vary by country; products may not be available in your geographic area.
Thermo Scientific™

PrioCHECK™ CSFV Antibody ELISA Kit, solid

Catalog number: 7610046
Have Questions?
Thermo Scientific™

PrioCHECK™ CSFV Antibody ELISA Kit, solid

Catalog number: 7610046
Have Questions?
Catalog Number
7610046
Unit Size
440 samples
Price (USD)
Catalog NumberUnit SizePrice (USD)
7610046440 samples
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The PrioCHECK® CSFV Antibody ELISA is a blocking ELISA for the detection of antibodies directed against CSFV in pigs. The test detects antibodies against high-, moderate-, and low-virulent CSFV strains early after infection. The PrioCHECK® CSFV Antibody ELISA uses two monoclonal antibodies (mAbs) to measure the binding of CSFV antibodies in the test sample directed against the E2 protein of CSFV. One mAb is bound to the test plate; the other, the detection antibody, is conjugated to an enzyme that generates a color signal. The signal is then measured and when no color is formed, the sample containing anti-CSFV antibodies that have competed for the available antigen is positive for CSFV.

Specifications

Assay Sensitivity
See product documentation
Immunoassay Kit Format
Other Format
Product Line
PrioCHECK™, Prionics™
Product Type
ELISA Kit
Protein Family
Other Proteins
Gene ID (Entrez)
Non-Gene
Gene Symbol
Non-Gene
Immunoassay Kit Grade
Solid
Target Kit Named
CSFV Antibody
Target Organism Class
Classical Swine Fever Virus (CSFV)
Target Species Validated
Porcine

Contents & Storage

5 plates

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text