Informational:Regulatory requirements vary by country; products may not be available in your geographic area.
Applied Biosystems™

PrioCHECK™ Bovine Salmonella Ab Strip Kit

Catalog number: 7610620
Have Questions?
Applied Biosystems™

PrioCHECK™ Bovine Salmonella Ab Strip Kit

Catalog number: 7610620
Have Questions?
Catalog Number
7610620
Unit Size
450 samples
Price (USD)
Catalog NumberUnit SizePrice (USD)
7610620450 samples
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Note: The name of this product has changed. It was previously called PRIOCHECK SALMONELLA AB BOV.

The PrioCHECK™ Salmonella Antibody bovine ELISAs are the first commercially available tests for monitoring and surveillance of Salmonella infections in cattle herds. The tests allow fast and versatile detection of anti-Salmonella antibodies in serum, plasma, and milk samples. Both ELISAs follow an easy 4-step protocol. Results are available within 150 min, making the tests highly suitable for screening and monitoring programs for bovine salmonellosis.

The PrioCHECK™ Salmonella Antibody ELISA Dublin detects S. dublin more specifically, but not exclusively. Some cross-reactivity with S. typhimurium can occur. Both tests are suitable for large-scale screening of serum, plasma, and (bulk) milk samples.

Test Procedure
Test samples are incubated in plates coated with the purified Salmonella antigen at room temperature. Plates are then washed and an anti-bovine antibody labeled with an enzyme that generates a color signal is added. The signal is measured and if color develops the sample is positive for anti-Salmonella antibodies. Color intensity is directly related to the amount of anti-Salmonella antibody present in the test sample.

Specifications

Assay Sensitivity
See product documentation
Immunoassay Kit Format
Other Format
Product Line
PrioCHECK™, Prionics™
Protein Family
Other Proteins
Target Species
Bovine
Gene ID (Entrez)
Non-Gene
Gene Symbol
Non-Gene
Immunoassay Kit Grade
Strip
Quantity
5 Plates
Target Kit Named
Salmonella Antibody
Target Organism Class
Salmonella
Target Species Validated
Bovine

Contents & Storage

5 Plates

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text