PrioCHECK™ S. dublin Ab Strip Kit
Regulatory requirements vary by country; products may not be available in your geographic area.
PrioCHECK™ S. dublin Ab Strip Kit
Applied Biosystems™

PrioCHECK™ S. dublin Ab Strip Kit

Note: The name of this product has changed. It was previously called PRIOCHECK SALMONELLA AB BOV DUBL. The PrioCHECK™ SalmonellaRead more
Have Questions?
Catalog NumberQuantity
7610640450 samples, 5 plates
Catalog number 7610640
Price (USD)
-
Quantity:
450 samples, 5 plates
Ask our AI about this Product
Note: The name of this product has changed. It was previously called PRIOCHECK SALMONELLA AB BOV DUBL.

The PrioCHECK™ Salmonella Antibody bovine ELISAs are the first commercially available tests for monitoring and surveillance of Salmonella infections in cattle herds. The tests allow fast and versatile detection of anti-Salmonella antibodies in serum, plasma, and milk samples. Both ELISAs follow an easy 4-step protocol. Results are available within 150 min, making the tests highly suitable for screening and monitoring programs for bovine salmonellosis.

The PrioCHECK™ Salmonella Antibody ELISA Dublin detects S. dublin more specifically, but not exclusively. Some cross-reactivity with S. typhimurium can occur. Both tests are suitable for large-scale screening of serum, plasma, and (bulk) milk samples.

Test Procedure
Test samples are incubated in plates coated with the purified Salmonella antigen at room temperature. Plates are then washed and an anti-bovine antibody labeled with an enzyme that generates a color signal is added. The signal is measured and if color develops the sample is positive for anti-Salmonella antibodies. Color intensity is directly related to the amount of anti-Salmonella antibody present in the test sample.
Specifications
Assay SensitivitySee product documentation
Immunoassay Kit FormatOther Format
Product LinePrioCHECK™, Prionics™
Protein FamilyOther Proteins
SpecificityS. dublin
Target SpeciesBovine
Gene ID (Entrez)Non-Gene
Gene SymbolNon-Gene
Immunoassay Kit GradeStrip
Quantity450 samples, 5 plates
Target Kit NamedSalmonella Antibody
Target Organism ClassSalmonella
Target Species ValidatedBovine
Unit SizeEach
Contents & Storage
5 Plates
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link