Specifications
Assay SensitivitySee product documentation
Immunoassay Kit FormatOther Format
Product LinePrioCHECK™, Prionics™
Protein FamilyOther Proteins
Sample TypeSerum
Target SpeciesBovine
Gene ID (Entrez)Non-Gene
Gene SymbolNon-Gene
Immunoassay Kit GradeStrip
Quantity460 Samples
Target Kit NamedBrucella Antibody 2.0
Target Organism ClassBrucella
Target Species ValidatedBovine, Goat, Sheep
Unit SizeEach