Informational:Regulatory requirements vary by country; products may not be available in your geographic area.
Applied Biosystems™

PrioCHECK™ Brucella Antibody 2.0 ELISA Kit, solid

Catalog number: 7610710
Have Questions?
Applied Biosystems™

PrioCHECK™ Brucella Antibody 2.0 ELISA Kit, solid

Catalog number: 7610710
Have Questions?
Catalog Number
7610710
Unit Size
2300 samples
Price (USD)
Catalog Number
Unit Size
460 samples
Price (USD)
Catalog NumberUnit SizePrice (USD)
76107102300 samples
7610700460 samples
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The PrioCHECK® Brucella Antibody 2.0 ELISA is an indirect ELISA for the detection of antibodies against Brucella abortus and Brucella melitensis in the serum and milk of cattle, sheep, and goats. The test meets EU requirements and OIE standards. The PrioCHECK® Brucella Antibody 2.0 ELISA procedure consists of 4 steps. In the first step, serum samples and milk samples are dispensed in the coated wells of a microtiterplate. Antibodies, directed against Brucella abortus and Brucella melitensis, present in the test sample, will bind to the antigen during incubation. The bound antibodies are detected using an anti-Ig monoclonal antibody, conjugated to an enzyme that generates a color signal. Color development will occur when specific antibodies against Brucella abortus or Brucella melitensis are present in the test sample. Color development indicates the presence of Brucella antibodies in the sample.

Specifications

Assay Sensitivity
See product documentation
Immunoassay Kit Format
Other Format
Product Line
PrioCHECK™, Prionics™
Protein Family
Other Proteins
Sample Type
Serum
Target Species
Bovine
Gene ID (Entrez)
Non-Gene
Gene Symbol
Non-Gene
Immunoassay Kit Grade
Solid
Quantity
2300 Samples
Target Kit Named
Brucella Antibody 2.0
Target Organism Class
Brucella
Target Species Validated
Bovine, Goat, Sheep

Contents & Storage

25 plates

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text