Thermo Scientific™
Imject™ Maleimide-Activated BSA
Catalog number: 77115
Have Questions?
Thermo Scientific™
Imject™ Maleimide-Activated BSA
Catalog number: 77115
Have Questions?
Catalog Number
77115
Unit Size
10 mg
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
2 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
7711510 mg
-
771162 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Imject Maleimide-Activated BSA enables conjugation of sulfhydryl-containing peptide antigens to immunogenic bovine serum albumin carrier protein for immunization and antibody production.
Features of Imject Maleimide-Activated BSA:
Optimized activation— specifications require that each lot is activated with 15 to 20 moles of sulhydryl-reactive maleimide per mole of BSA, ensuring consistently high levels of conjugation
Sulfhydryl-reactive— engineered for conjugating cysteine-containing peptide antigens to make effective immunogens for high-titer antibody production
Simple, rapid procedure— complete the hapten-carrier conjugation reaction in two hours to yield sufficient immunogen for several injections and booster immunizations of host animals
High-quality carrier protein— pure BSA (Fraction V) is activated using high-quality Thermo Scientific Sulfo-SMCC crosslinker and formulated to ensure high stability and consistent performance
Immunogenic and soluble— moderately immunogenic and highly soluble, BSA is useful for preparation of the primary immunogen or as an irrelevant hapten-carrier for antibody screening and immunoassays after using KLH as the carrier protein to generate the immune response against the hapten
Purified, carrier-grade BSA is rendered sulfhydryl-reactive by chemical modification with Sulfo-SMCC crosslinker, and the resulting maleimide-activated protein is then stabilized and lyophilized in phosphate buffer. Maleimide groups form covalent crosslinks with sulfhydryl (-SH) moieties on cysteine residues of peptides and other thiol-containing haptens. Several antigen molecules can be conjugated to each BSA molecule to increase their immunogenicity and likelihood of eliciting immunized animals to produce high-titer antisera to the epitopes of interest. Select from among several package sizes of activated BSA protein, or choose the complete conjugation kit that includes optimized buffers and trouble-free spin desalting columns.
Carrier proteins are large, complex molecules capable of stimulating an immune response upon injection. Successful production of antibodies specific to small antigens (i.e., peptides or drug compounds) requires that these haptens be covalently conjugated to a larger, more complex molecule (usually a protein) to make them immunogenic. Carrier proteins are chosen based on immunogenicity, solubility, and whether an adequate level of hapten-carrier conjugation can be achieved.
Bovine serum albumin (BSA) is a convenient protein for a variety of uses in the laboratory because, like most abundant plasma proteins, it is very stable and soluble. Numerous carboxyl groups account for the net negative charge (pI 5.1) of BSA and its high solubility (compare to positively-charged, cationized BSA). In addition, the 67kDa protein is sufficiently large and complex to be fully immunogenic. Consequently, BSA is a popular carrier protein for conjugation to haptens and other weak antigens to make them more immunogenic for the purpose of antibody production. If not used to prepare the primary immunogen, BSA is a useful carrier protein to make antigens more amenable to analysis (e.g., screening assays involving plate coating) of antibodies that were produced using KLH or another more immunogenic carrier protein.
More Product Data
Carrier protein activation and conjugation data for immunogen preparation
Related Products
Imject™ Maleimide-Activated BSA Spin Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Description
Imject Maleimide Activated Bovine Serum Albumin
Quantity
10 mg
Product Line
Imject™
Contents & Storage
Upon receipt store product at 4°C. Product is shipped at ambient temperature.