1x1 image pixel for data collection
Imject™ 马来酰亚胺偶联缓冲液
Imject™ 马来酰亚胺偶联缓冲液
Thermo Scientific™

Imject™ 马来酰亚胺偶联缓冲液

Thermo Scientific Imject Maleimide Conjugation Buffer, formulation: 0.083M sodium phosphate, 0.1M EDTA, 0.9M NaCl, 0.1M sorbitol and 0.02% sodium azide;了解更多信息
Have Questions?
货号数量
7716430 mL
货号 77164
价格(CNY)
973.00
Each
下单后预计有货日期 21-Jul-2025
添加至购物车
数量:
30 mL
价格(CNY)
973.00
Each
添加至购物车
Thermo Scientific Imject Maleimide Conjugation Buffer, formulation: 0.083M sodium phosphate, 0.1M EDTA, 0.9M NaCl, 0.1M sorbitol and 0.02% sodium azide; pH 7.2. 30 mL of this buffer is suffienct for dissolving up to 60 mg of sulfhydryl-peptide for maleimide conjugation with carrier proteins.

More Product Data
Carrier protein activation and conjugation data for immunogen preparation

Related Products
Imject™ Maleimide-Activated mcKLH
Imject™ Maleimide-Activated mcKLH Spin Kit
Imject™ Maleimide PEGylated mcKLH
仅供科研使用。不可用于诊断程序。
规格
描述Imject 马来酰亚胺交联缓冲液
数量30 mL
产品线Imject™
Unit SizeEach
内容与储存
储存在原容器中,置于干燥、阴凉且通风良好的区域内,避免阳光直照。

图表

文件和下载

证书

批号Certificate TypeDateCatalog Number(s)
3215988Certificate of Analysis2025年5月29日77164
ZH393800Certificate of Analysis2024年10月15日77164
YG380829Certificate of Analysis2023年8月23日77164
YB363968Certificate of Analysis2023年2月28日77164
XI355140Certificate of Analysis2022年10月15日77164
显示 5 条结果, 在上面搜索特定证书

安全数据表

其他推荐产品



分享货号、产品名称或链接

1x1 image pixel for data collection