Thermo Scientific™

Imject™ Maleimide-Activated BSA Spin Kit

Catalog number: 77667
Have Questions?
Thermo Scientific™

Imject™ Maleimide-Activated BSA Spin Kit

Catalog number: 77667
Have Questions?
Catalog Number
77667
Unit Size
5 x 2 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
776675 x 2 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific Imject Maleimide-Activated BSA Spin Kit contains stabilized, lyophilized Maleimide-Activated BSA (in PBS with stabilizer) and accessory components to easily prepare ready-to-use immunogens for immunization. Our Imject Maleimide-Activated BSA enables conjugation of sulfhydryl-containing peptide antigens to immunogenic bovine serum albumin carrier protein for immunization and antibody production.

Features of Imject Maleimide-Activated BSA:

Optimized activation— specifications require that each lot is activated with 15 to 20 moles of sulhydryl-reactive maleimide per mole of BSA, ensuring consistently high levels of conjugation
Sulfhydryl-reactive— engineered for conjugating cysteine-containing peptide antigens to make effective immunogens for high-titer antibody production
Simple, rapid procedure— complete the hapten-carrier conjugation reaction in two hours to yield sufficient immunogen for several injections and booster immunizations of host animals
High-quality carrier protein— pure BSA (Fraction V) is activated using high-quality Thermo Scientific Sulfo-SMCC crosslinker and formulated to ensure high stability and consistent performance
Immunogenic and soluble— moderately immunogenic and highly soluble, BSA is useful for preparation of the primary immunogen or as an irrelevant hapten-carrier for antibody screening and immunoassays after using KLH as the carrier protein to generate the immune response against the hapten

Purified, carrier-grade BSA is rendered sulfhydryl-reactive by chemical modification with Sulfo-SMCC crosslinker, and the resulting maleimide-activated protein is then stabilized and lyophilized in phosphate buffer. Maleimide groups form covalent crosslinks with sulfhydryl (-SH) moieties on cysteine residues of peptides and other thiol-containing haptens. Several antigen molecules can be conjugated to each BSA molecule to increase their immunogenicity and likelihood of eliciting immunized animals to produce high-titer antisera to the epitopes of interest. Select from among several package sizes of activated BSA protein, or choose the complete conjugation kit that includes optimized buffers and trouble-free spin desalting columns.

Carrier proteins are large, complex molecules capable of stimulating an immune response upon injection. Successful production of antibodies specific to small antigens (i.e., peptides or drug compounds) requires that these haptens be covalently conjugated to a larger, more complex molecule (usually a protein) to make them immunogenic. Carrier proteins are chosen based on immunogenicity, solubility, and whether an adequate level of hapten-carrier conjugation can be achieved.

Bovine serum albumin (BSA) is a convenient protein for a variety of uses in the laboratory because, like most abundant plasma proteins, it is very stable and soluble. Numerous carboxyl groups account for the net negative charge (pI 5.1) of BSA and its high solubility (compare to positively-charged, cationized BSA). In addition, the 67kDa protein is sufficiently large and complex to be fully immunogenic. Consequently, BSA is a popular carrier protein for conjugation to haptens and other weak antigens to make them more immunogenic for the purpose of antibody production. If not used to prepare the primary immunogen, BSA is a useful carrier protein to make antigens more amenable to analysis (e.g., screening assays involving plate coating) of antibodies that were produced using KLH or another more immunogenic carrier protein.

More Product Data
Carrier protein activation and conjugation data for immunogen preparation

Related Products
Imject™ Maleimide-Activated BSA
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Description
Imject Maleimide-Activated BSA Spin Kit
Format
Kit
Quantity
5 x 2 mg Kit
Product Line
Imject™

Contents & Storage

Sufficient For: 5 conjugation reactions and complete immunogen preparations, each with up to 2 mg sulfhydryl-containing peptide
• Imject Maleimide-Activated BSA, 5 x 2 mg
• Imject Maleimide Conjugation Buffer, 30 mL
• Imject Purification Buffer Salts, 5 x 0.84 g
• Zeba Spin Desalting Columns, 7K MWCO, 2 mL, 5 columns

Store the Maleimide-Activated blue Carrier Protein at -20°C. Store remaining components at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text