Imject™ EDC BSA Spin Kit
Product Image
Thermo Scientific™

Imject™ EDC BSA Spin Kit

The Thermo Scientific Imject EDC BSA Spin Kit is a package of purified bovine serum albumin carrier protein that enableRead more
Have Questions?
Catalog NumberQuantity
776725 x 2 mg
Catalog number 77672
Price (USD)
716.00
Each
-
Add to cart
Quantity:
5 x 2 mg
Price (USD)
716.00
Each
Add to cart
The Thermo Scientific Imject EDC BSA Spin Kit is a package of purified bovine serum albumin carrier protein that enable simple preparation of effective immunogens with amine- or carboxyl-peptide antigens.

BSA is a popular immunogenic carrier protein used for preparation of peptide antigens for immunization and antibody production. Preparations in MES buffer are optimized for peptide-carrier conjugation via the carboxyl-reactive carbodiimide crosslinker called EDC (or EDAC). The Imject EDC Spin Kit includes ready-to-use units of BSA carrier protein, optimized reaction buffers and trouble-free spin desalting columns for immunogen preparation using the EDC method.

Features of the Imject EDC BSA Spin Kit:

Convenient kit—contains lyophilized BSA (in MES buffer), EDC crosslinker and accessories components to easily prepare ready-to-use immunogens for injection
High-yield conjugation—each molecule of 59 lysine residues, 30 to 35 of which have primary amines that are capable of reacting with a conjugation reagent
Validated quality—purified BSA (Fraction V) is ready for use in glutaraldehyde, NHS-ester or EDC crosslinking methods
Compatible solubility—numerous carboxyl groups account for the net negative charge (pI 5.1) of BSA and its high solubility, allowing its use in a broad range of conjugation conditions
Immunogenic—at 67 kDa, BSA is sufficiently large to function as the primary immunogen or it can be used as an irrelevant protein carrier for antibody screening and immunoassays after using KLH as the carrier protein to generate the immune response against the hapten

Carrier proteins are large, complex molecules capable of stimulating an immune response upon injection. Successful production of antibodies specific to small antigens (i.e., peptides or drug compounds) requires that these haptens be covalently conjugated to a larger, more complex molecule (usually a protein) to make them immunogenic. Carrier proteins are chosen based on immunogenicity, solubility, and whether adequate hapten-carrier conjugation can be achieved. Imject BSA is very soluble in aqueous buffers and contains numerous primary amines and carboxyl groups that can be targeted for conjugation with glutaraldehyde, NHS esters, EDC and other crosslinking reagents. Imject BSA is purified and lyophilized in buffers that optimize its stability and solubility for hapten conjugation.

Bovine serum albumin (BSA) is a convenient protein for a variety of uses in the laboratory because, like most abundant plasma proteins, it is very stable and soluble. In addition, the 67 kDa protein is sufficiently large and complex to be fully immunogenic. Consequently, BSA is a popular carrier protein for conjugation to haptens and other weak antigens to make them more immunogenic for the purpose of antibody production. It also provides an independent hapten-carrier protein for microplate and other assay techniques required to screen antibodies produced using KLH or another more immunogenic carrier.

The carbodiimide crosslinker EDC conjugates carboxyl-containing haptens (e.g., C-terminus of peptide antigens) to BSA carrier protein. This method of immunogen preparation is ideal for peptide antigens with few or no aspartic and glutamic acid residues (carboxylates) and lysine residues (primary amines) within the central portion of the primary sequence. Because peptides contain both carboxylate and amines, EDC conjugation results in their becoming variously polymerized and randomly oriented in their linkage to the carrier protein. Typically, this results in a high level of antigen loading on the carrier protein as well as presentation in all possible orientations for antibody production. However, important (desired) epitopes within the antigen peptide sequence may be blocked by EDC-mediated conjugation if those regions contain primary amines (lysine residues) or carboxylates (aspartic and glutamic acid residues). In these cases, either use a homobifunctional amine-reactive crosslinker with the purified BSA in phosphate buffer or synthesize the peptide with a unique terminal cysteine and use a kit with Malemide-Activated BSA to prepare the carrier protein conjugate.

More Product Data
Carrier protein activation and conjugation data for immunogen preparation

Related Products
Imject™ BSA (in PBS)
Imject™ BSA (in MES Buffer)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionImject EDC BSA Spin Kit
FormatKit
Quantity5 x 2 mg
Product LineImject™
Unit SizeEach
Contents & Storage
Sufficient For: 5 EDC conjugation reactions and complete immunogen preparations, each with up to 2 mg peptide
• Imject BSA (in MES buffer), 5 x 2 mg
• Imject EDC Conjugation Buffer, 30 mL
• EDC Crosslinker, 5 x 10 mg
• Imject Purification Buffer Salts, 5 x 0.84 g
• Zeba Spin Desalting Columns, 7K MWCO, 2 mL, 5 columns

Store at 4°C.

Frequently asked questions (FAQs)

What do I need to consider when designing a peptide for antibody production?

We recommend that hydrophobic residues comprise 50% or less of all the residues in your sequence. Make sure there is at least one charged residue for every five amino acids: this is generally known to enhance the solubility of the peptide. Peptides (compared to polypeptides, which fold and bury the hydrophobic amino acids) are too small to fold, so just a few hydrophobic amino acids may leave them insoluble.

Other guidelines include:
Peptides containing multiple Cys, Met, and Trp can be hard to synthesize.
Some sequences are problematic in solid-phase peptide synthesis or cleavage and are best avoided; these include Asp-Pro sequences or stretches of amino acids that require bulky protecting groups on their side chains during synthesis.
Glycine is often good for antigenicity as it has only a hydrogen side chain; this allows for complete rotation.
Please note that we offer a Custom Antibody Production service (https://www.thermofisher.com/us/en/home/life-science/antibodies/custom-antibodies/custom-antibody-production.html) that includes the use of our proprietary Antigen Profiler and Antigen Preparation tool (https://www.thermofisher.com/us/en/home/life-science/antibodies/custom-antibodies/custom-antibody-production/antigen-profiler-antigen-preparation.html).

What is the optimal peptide length for antibody production?

Most peptide antigens range in length from 12 to 16 residues and are relatively easy to synthesize. Peptides of 9 residues or shorter have been effective antigens for antibody production, but peptides longer than 16 amino acids may contain several epitopes and form secondary structures. Peptides in excess of 18 residues begin to present more synthetic challenges. Before you synthesize your peptide, we recommend doing a BLASTP search using your peptide sequence. This is to make sure that the peptide is not homologous or identical to a sequence in a completely unrelated protein in the host animal.

Please note that we offer a Custom Antibody Production service (https://www.thermofisher.com/us/en/home/life-science/antibodies/custom-antibodies/custom-antibody-production.html) that includes the use of our proprietary Antigen Profiler and Antigen Preparation tool (https://www.thermofisher.com/us/en/home/life-science/antibodies/custom-antibodies/custom-antibody-production/antigen-profiler-antigen-preparation.html).

What happens if antibodies are stored improperly?

Improper storage of antibodies can lead to:
- Degradation: Loss of activity and specificity
- Aggregation: Formation of precipitates or clumps
- Reduced performance: Poor results in assays or experiments

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.

Is your EDAC compound the same as your EDC compound?

Yes, both acronyms are used for the same compound. The chemical name for EDC is 1-ethyl-3-(3-dimethylaminopropyl)-carbodiimide and the chemical name for EDAC is N-3-(3-dimethylaminopropyl)-N-ethyl-carbodiimide. Both of these compounds are structurally equivalent.

What do antibody pair kits contain, how many samples can I process, and how can I find a list of antibody pair kits by target?

Antibody pair kits contain capture antibody, detection antibody, recombinant standard and HRP conjugate. Each contains enough reagents to process forty 96-well plates. A list of Antibody Pair Kits (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/elisa/antibody-pair-kits.html) is available by target.

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link