1x1 image pixel for data collection
DynaGreen™ Protein A Magnetic Beads
DynaGreen™ Protein A Magnetic Beads
Invitrogen™

DynaGreen™ Protein A Magnetic Beads

Green features
DynaGreen Protein A Magnetic Beads are intended for use in immunoprecipitation (IP) applications while also being an environmentally sustainable choice.Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
80103G25 mL, 1000 Reactions
80101G0.5 mL, 20 Reactions
80102G3 mL, 120 Reactions
Catalog number 80103G
Price (USD)
4,544.65
Online Exclusive
4,680.00
Save 135.35 (3%)
Each
In stock
Add to cart
Quantity:
25 mL, 1000 Reactions
Price (USD)
4,544.65
Online Exclusive
4,680.00
Save 135.35 (3%)
Each
Add to cart

DynaGreen Protein A Magnetic Beads are intended for use in immunoprecipitation (IP) applications while also being an environmentally sustainable choice. They are microplastic-free submicron magnetic beads that provide reproducible isolation of target proteins in a simple and efficient workflow, which can be performed manually or automated on KingFisher™ instruments.

Benefits of DynaGreen Protein A Magnetic Beads include:

  • High performance—reproducible IP with high-yield and low non-specific binding
  • Reduced environmental impact—non-microplastic bead core and holistic sustainable product design
  • Flexible—direct and indirect immunoprecipitation helps ensure isolation of non-abundant proteins
  • Automation ready—easy automation for high-throughput needs on the Kingfisher Sample Purification Systems

Simple, gentle, and efficient manual separation

DynaGreen Protein A Magnetic Beads are microplastic-free superparamagnetic beads with recombinant protein A (∼45 kDa) covalently coupled to the surface. They were developed for convenient IP of target proteins when the IP antibody interacts strongly with Protein A (see subclass specificity in the manual). The submicron bead size (∼250 nm) provides low sedimentation and a large target capture surface area. The method is a simple bind-incubate-elute protocol with no pre-clearing required. This results in an efficient high-yield isolation of target protein by direct or indirect immunoprecipitation in less than 80 minutes.

The magnetic separation technology used by DynaGreen Protein A Magnetic Beads is rapid and gentle, thereby reducing physical stress to the target proteins. Furthermore, the non-porous nature of the beads lowers the risk of antibodies being trapped and becoming inaccessible. This helps to minimize the amount of antibody required to produce accurate, and reproducible results. The yield and purity of the isolated target protein are highly compatible for use in western blotting and mass spectrometry. The product manual below includes a list of required reagents, materials, and details of compatible buffer recipes.

Rapid and reproducible automated workflow

The DynaGreen magnetic beads are ideal for high-throughput enrichment. This process can be automated using any of our KingFisher sample purification systems or liquid handlers.

The automated protocol can be performed in just 40 minutes with up to 96 samples per run, which helps reduce hands-on time while maintaining the same reproducible high target yield and low non-specific binding compared to the manual protocol.

A certified choice for environmentally sustainable research

Microplastic-free DynaGreen Protein A Magnetic Beads are designed for both high performance in immunoprecipitation workflows and lower environmental impact. They provide exceptional purity and high yield of target proteins compared to other magnetic beads in both direct and indirect immunoprecipitation workflows. At the same time, the product boasts an extensive list of third-party ACT-label-certified sustainable product design elements by incorporating principles of green chemistry and green engineering.

Choosing DynaGreen Protein A Magnetic Beads can help reduce the environmental impact of research without compromising on results.

Learn more about our DynaGreen magnetic beads and KingFisher Sample Purification Systems:

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Certifications/ComplianceISO9001 and ISO13485
ColorBrown
Concentration20 mg/mL
DescriptionRecombinant Protein A covalently bound to DynaGreen magnetic beads
Diameter (Metric)250 nm
For Use With (Application)Immunoprecipitation
For Use With (Equipment)KingFisher™ Sample Purification Systems, DynaMag™ Magnets
Green FeaturesLess hazardous, less waste, sustainable packaging
High-throughput CompatibilityHigh-throughput Compatible
Ligand TypeRecombinant Protein A
MaterialIron Oxide
Purity or Quality GradeFor Research Use Only
Quantity25 mL, 1000 Reactions
Shelf Life12 months from date of manufacture
Shipping ConditionAmbient temperature
Sufficient For1000 Tests
TargetProteins
Product LineDynaGreen™
TypeProtein A Magnetic Bead
Unit SizeEach
Contents & Storage
DynaGreen Protein A Magnetic Beads are supplied in phosphate buffer, pH 7.4, with a biodegradable surfactant and a preservative. Store at 2–8°C.

Frequently asked questions (FAQs)

What can I do if the antibody from the DynaGreen magnetic beads is coming off the beads during the elution step?

The antibody will come off the beads during elution if the antibody is not cross-linked to the beads. The antibody is attached to the Protein A/G through affinity binding, thus when you break one of the affinity bindings, the other affinity bindings will break as well.

Find additional tips, troubleshooting help, and resources within our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

Do you have any tips to offer if we are experiencing non-specific protein binding to DynaGreen magnetic beads or proteins trapped in the protein-antibody-antigen immunocomplex?

Here are some tips:
- Perform a pre-clearing procedure before doing immunoprecipitation (IP) by adding the beads to the sample without the antibody bound. After this pre-clearing step, continue with the IP protocol using the antibody-bound beads.
- Increase the washing time and increase the number of washing steps after IP.
- Increase the stringency of the washes, e.g., wash with a buffer containing a higher concentration of detergent or salt.
- Try the indirect IP technique (add antibody to the sample first, and then add the beads), change to another DynaGreen product (e.g., switch from DynaGreen Protein A/G Magnetic Beads to DynaGreen CaptureSelect Anti IgG-Fc Magnetic Beads), or try Dynabeads Protein A or Dynabeads Protein G for Immunoprecipitation.

Find additional tips, troubleshooting help, and resources within our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

When using DynaGreen magnetic beads, how can we avoid sample degradation by proteases?

We recommend including additional protease inhibitors in the lysis and wash buffers and keeping the sample cold at all times.

Find additional tips, troubleshooting help, and resources within our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

How can I improve the elution of the target from DynaGreen magnetic beads using a low pH buffer?

The addition of a small amount of detergent (eg., 0.05% of Tween 20) in the wash buffer, before elution, can improve the elution of the target. Also, it is important to fully remove the wash buffer before adding the elution buffer, in order to maintain the low pH conditions required for elution.

Find additional tips, troubleshooting help, and resources within our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

Are DynaGreen magnetic beads sterile?

Per definition, DynaGreen magnetic beads are not sterile, but all the buffers used in the manufacturing process are autoclaved and a biocide called Acticide is added to the product to help keep it free of any type of growth.

Find additional tips, troubleshooting help, and resources within our Protein Immunoprecipitation (IP), Co-Immunoprecipitation (Co-IP), and Pulldown Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection