Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Accomodate a variety of applications with uniquely designed, innovative Thermo Scientific™ Matrix Reagent Reservoirs. These sterile reservoirs offer an extra-wide base to add rigidity and stability, helping to avoid spills. The 25mL divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other; maximizing recovery if using only a single channel and a small amount of reagent.
Color | White |
Capacity (Metric) | 100 mL |
Material | Polystyrene |
Sterility | Sterile |
Type | Reagent Reservoir |
Packaging | 50 Individually wrapped |
Unit Size | Pack of 50 |
Color | White |
Capacity (Metric) | 75 mL |
Material | Polypropylene |
Sterility | Non-sterile |
Type | Reagent Reservoir |
No. per Case | 10 |
No. per Pack | 10 |
Packaging | 10 pcs /bag |
Autoclavable | Autoclavable |
Unit Size | Case of 10 |
Color | White |
Capacity (Metric) | 75 mL |
Material | Polypropylene |
Sterility | Non-sterile |
Type | Reservoir Lid |
No. per Case | 10 |
No. per Pack | 10 |
Packaging | 10 pcs /bag |
Unit Size | Case of 10 |
Color | White |
Capacity (Metric) | 100 mL |
Material | Polystyrene |
Sterility | Sterile |
Type | Reagent Reservoir |
Packaging | 10 Bags of 10 |
Unit Size | Pack of 100 |
Capacity (Metric) | 25 mL |
Material | Polystyrene |
Sterility | Sterile |
Type | Reagent Reservoir |
Packaging | 10 Bags of 10 |
Unit Size | Each |
Color | White |
Capacity (Metric) | 25 mL |
Material | Polystyrene |
Sterility | Sterile |
Type | Reagent Reservoir |
Packaging | 50 Individually wrapped |
Unit Size | Pack of 50 |
Color | White |
Capacity (Metric) | 25 mL |
Material | Polystyrene |
Sterility | Sterile |
Type | Reagent Reservoir with Divider |
Packaging | 10 Bags of 10 |
Unit Size | Pack of 100 |
Capacity (Metric) | 25 mL |
Material | Polystyrene |
Sterility | Sterile |
Type | Reagent Reservoir with Divider |
Packaging | 50 Individually wrapped |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Capacity (Metric) | Material | Packaging | Type | Price (JPY) | Availability | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
8086 | Pack of 50 | 100 mL | Polystyrene | 50 Individually wrapped | Reagent Reservoir | Price: 16,000 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
8075 | Case of 10 | 75 mL | Polypropylene | 10 pcs /bag | Reagent Reservoir | Price: 13,200 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
8076 | Case of 10 | 75 mL | Polypropylene | 10 pcs /bag | Reservoir Lid | Price: 5,900 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
8085 | Pack of 100 | 100 mL | Polystyrene | 10 Bags of 10 | Reagent Reservoir | Price: 16,000 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
8093 | Each | 25 mL | Polystyrene | 10 Bags of 10 | Reagent Reservoir | Request A Quote | *** | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
8094 | Pack of 50 | 25 mL | Polystyrene | 50 Individually wrapped | Reagent Reservoir | Price: 16,000 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
8095 | Pack of 100 | 25 mL | Polystyrene | 10 Bags of 10 | Reagent Reservoir with Divider | Price: 16,000 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
8096 | Each | 25 mL | Polystyrene | 50 Individually wrapped | Reagent Reservoir with Divider | Request A Quote | *** | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|