Orion™ pH Electrode Storage Solutions
Product Image
Thermo Scientific™

Orion™ pH Electrode Storage Solutions

Restore proper electrode performance and prolong electrode life using these pH electrode storage solutions.
Have Questions?
Ansicht ändernbuttonViewtableView
KatalognummerZur Verwendung mit (Geräte)Menge
810001TS
auch als 810001 bezeichnet
pH-Elektrode475 ml
910001pH-Elektrode475 ml
910060pH-Elektrode60 ml
9100CBpH-Elektrode19 L
Katalognummer 810001TS
auch als 810001 bezeichnet
Preis (EUR)
75,70
Each
-
Zum Warenkorb hinzufügen
Zur Verwendung mit (Geräte):
pH-Elektrode
Menge:
475 ml
Recurring order eligible. Learn more »
Preis (EUR)
75,70
Each
Zum Warenkorb hinzufügen
Ask our AI about this Product

Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH Electrode Storage Solutions in ready to use bottles.

Storage Solutions:

  • Storage solution ensures that the pH electrode works properly after prolonged storage, overnight storage and between samples


Warranty: Out of box

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
BeschreibungAufbewahrungslösung für ROSS pH-Elektroden, 475 ml-Flasche (1 pint)
Art der FülllösungAufbewahrungslösung
VerpackungFlasche
Menge475 ml
Zur Verwendung mit (Geräte)pH-Elektrode
TypLagerungslösung für pH-Elektroden
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Do I need to buy the AutoLys M TubeLifter (Cat. No. A37956) or AutoLys M Tube Pliers (Cat. No. A38261) to use the AutoLys M Tubes (Cat. No. A38738)?
Why is no RNase digestion step required when I only want to isolate DNA from FFPE samples using the MagMAX FFPE DNA/RNA Ultra Kit?
Can the buffers of the MagMAX FFPE DNA/RNA Ultra Kit be used for the MagMAX FFPE Total Nucleic Acid Isolation Kit and vice versa?
I can see on your website that you offer several MagMAX kits for FFPE samples. Which kit do you generally recommend?
How can I increase my chances of successful extraction of FFPE samples in terms of DNA quality and yield?
+Show more FAQs for this product
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Videos

Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
 
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Customers who viewed this item also viewed



      Dokumente und Downloads

      Zertifikate

      Chargen-Nr.Certificate TypeDateCatalog Number(s)
      0004508678Certificate of Analysis19. Juni 2025910001
      0004505952Certificate of Analysis22. Mai 2025910001
      SDV3Certificate of Analysis21. Mai 2025910001
      SDV2Certificate of Analysis21. Mai 2025910001
      0004497693Certificate of Analysis12. Mai 2025910001
      5 Ergebnisse angezeigt, oben nach einem bestimmten Zertifikat suchen

      Sicherheitsdatenblätter

      Geben Sie die Katalognummer, den Namen oder Link frei.

      1x1 image pixel for data collection