CaptureSelect™ AAVX Ligand Leakage ELISA Kit
CaptureSelect™ AAVX Ligand Leakage ELISA Kit
Thermo Scientific™

CaptureSelect™ AAVX Ligand Leakage ELISA Kit

The Thermo Scientific™ CaptureSelect™ AAVX Ligand Leakage ELISA Kit is a ready-to-use kit specifically designed to detect possible leached ligand present in adeno-associated viruses (AAV) purified using POROS CaptureSelect AAVX Affinity Resin.
Have Questions?
Catalog NumberQuantity
81135220001 kit
Catalog number 8113522000
Price (USD)
1,520.00
Each
In stock
Add to cart
Quantity:
1 kit
Price (USD)
1,520.00
Each
Add to cart

CaptureSelect AAVX Ligand Leakage ELISA Kit is a ready-to-use kit specifically designed to detect possible leached ligand present in adeno-associated viruses (AAV) purified using POROS CaptureSelect AAVX Affinity Resin. Although the ligand is covalently coupled, it is possible for the ligand to leach and co-elute with the purified AAV. This fully assembled kit consists of a pre-coated anti-AAVX ligand strip plate, buffers, and optimal reaction solutions, enabling a robust ELISA assay to measure potential ligand leakage.

Key features:

  • Reduced assay operating time, due to the pre-coating and pre-blocking of the strip plate
  • Quick and easy-to-set-up assay procedure
  • Sensitive and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL

The CaptureSelect AAVX Ligand Leakage ELISA Kit can be used as a tool to aid in optimal purification process development and routine quality control of in-process streams as well as final product. The kit is manufactured in an ISO-certified manufacturing site and is ISO 9001-certified.

Specifications
Assay96-well
Detection MethodELISA
For Use With (Application)Detection of possible leached ligand
Format96-well plate: 8-well strip plate, assembled 12 strips per plate
Quantity1 kit
Shipping Condition2°C to 8°C
TargetAnti AAVX ligand
Product LineCaptureSelect™
TypeELISA Kit
Unit SizeEach
Contents & Storage
• Pre-coated anti-AAVX ligand strip plate
• Wash/dilutionbuffer (10X)
• AAV standard/coating and biotin reagents
• Streptavidin-HRP conjugate reagent
• TMB substrate
• Stop solution

Store at 2–8°C.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link