Orion™ 8135BN ROSS™ Combination Flat Surface pH Electrode
Orion™ 8135BN ROSS™ Combination Flat Surface pH Electrode
Actual product may vary
Thermo Scientific™

Orion™ 8135BN ROSS™ Combination Flat Surface pH Electrode

Get stable readings to 0.01 pH in less than 30 seconds with this combination flat surface pH electrode.
Have Questions?
Catalog number 8135BN
Price (USD)/ Each
609.62
Special Offer
Online exclusive
Ends: 31-Dec-2024
713.00 
Save 103.38 (15%)
-
Add to cart
Connector Types:
BNC
Price (USD)/ Each
609.62
Special Offer
Online exclusive
Ends: 31-Dec-2024
713.00 
Save 103.38 (15%)
Add to cart
Orion™ 8135BN ROSS™ Combination Flat Surface pH Electrode
Catalog number8135BN
Price (USD)/ Each
609.62
Special Offer
Online exclusive
Ends: 31-Dec-2024
713.00 
Save 103.38 (15%)
-
Add to cart

Get stable readings to 0.01 pH in less than 30 seconds with the Thermo Scientific™ Orion™ 8135BN ROSS™ Combination Flat Surface pH Electrode. This instrument provides results three to five times more precise than those obtained with conventional electrodes.

The 8135BN ROSS Combination Flat Surface pH Electrode measures pH of soft, moist surfaces — both solid and semi-solid —such as agar gel plates, meats, bread dough and similar samples. With drift less than 0.002 pH per day and minimized restandardization, this electrode works in extreme cases of samples varying from one another by 50°C or more.

  • Material: Epoxy body
  • pH range: 0 to 14
  • Temperature range: 0° to 100°C
  • Internal reference: ROSS
  • Junction: Glass fiber
  • Dimensions: 120 × 12mm
  • pH precision: 0.01
  • Fill solution: Orion 810007
  • Cap diameter: 16mm
  • Small samples are available in micro sample dish
  • Flat surface tip

Includes: One 60 ml bottle of ROSS fill Solution, 810007 and spout cap


Warranty: 12 months from date of purchase by Buyer or 18 months from date of shipment from Seller, whichever is earlier

Specifications
DescriptionWith BNC connector
Connector TypesBNC
JunctionGlass Fiber
MaterialEpoxy Body
pH Range0 to 14
Temperature (Metric) Operating0°C to 100°C
TypeCombination Flat Surface pH Electrode, BNC connector
Drift<0.002pH/day
Filling Solution Type3M KCl
For Use With (Equipment)Standard pH Meter, BNC connector
Internal ReferenceROSS
Precision0.01
Storage RequirementsShort-term Storage (up to one week): Soak the electrode in ROSS' pH electrode storage solution, Cat. No. 810001, or standard pH electrode storage solution, Cat. No. 910001. For enhanced electrode performance, use ROSS' pH electrode storage solution. Long-term Storage (more than one week): Fill the reference chamber and securely cover the fill hole. Cover the sensing bulb and reference junction with the protective cap, sleeve or storage bottle containing a few drops of storage solution. Before returning the electrode to use, prepare it as a new electrode.
Temperature (English) Operating32°F to 212°F
Length (English) Cable39 in.
Length (Metric) Cable1 m
Diameter (English) Body0.47 in.
Diameter (Metric) Body12 mm
Diameter (Metric) Cap16 mm
Unit SizeEach