Thermo Scientific™

Sigma-Aldrich™ Enzymes and Coenzymes FT-IR Spectral Library

Catalog number: 834-005801
Have Questions?
Thermo Scientific™

Sigma-Aldrich™ Enzymes and Coenzymes FT-IR Spectral Library

Catalog number: 834-005801
Have Questions?
Use this material-specific subset of the larger Sigma Biochemical Condensed Phase Library for identification related to enzymes, coenzymes and enzyme substrates. The Thermo Scientific™ Sigma Enzymes, Coenzymes FT-IR Spectral Library helps eliminate guesswork and create greater efficiencies in your lab. Our comprehensive libraries feature fully-reviewed, high-quality spectroscopy databases, enabling confident compound identification for a variety of environmental, forensic, pharmaceutical and industrial applications.
 
Catalog Number
834-005801
Unit Size
Each
Description
Enzymes, Coenzymes FT-IR Spectral Library
Price (USD)
Full specifications
DescriptionEnzymes, Coenzymes FT-IR Spectral Library
For Use With (Application)FT-IR
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
834-005801Full specifications
EachEnzymes, Coenzymes FT-IR Spectral LibraryRequest A Quote
DescriptionEnzymes, Coenzymes FT-IR Spectral Library
For Use With (Application)FT-IR
Unit SizeEach
Showing 1 of 1
  • Spectra acquired by Sigma-Aldrich Co.™ and examined and processed by Thermo Fisher Scientific.
  • The spectra include compound name, molecular formula, CAS (Chemical Abstract Service) registry number and Sigma catalog number.
  • Contains 485 spectra
  • Data format: Deresolved
About Spectral Libraries
Spectral library searching has long been an extraordinarily valuable tool in the analytical chemist's toolbox. Our extensive series of spectral libraries includes information specific to application, industry or spectral technique.
Spectral library searching based on digital matching of spectral signatures is a very effective way of performing qualitative identification of compounds. Because FT-IR and Raman spectroscopic signatures are based on molecular functional groups, even if the exact compound is not present in the libraries, the list of matching compounds from a spectral library search can often lead to identification, or at least classification, of the material.
Thermo Scientific libraries span applications for Hazmat, Forensics, Food, Polymers and Organic Chemistry.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text