Coatings Technology FT-IR Spectral Library
Coatings Technology FT-IR Spectral Library
Actual product may vary
Thermo Scientific™

Coatings Technology FT-IR Spectral Library

Analyze of a wide variety of common coating materials with this valueable spectral library and associated handbook.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
834-010901Spectral Library
834-010901MSpectral Library, Member Price
Catalog number 834-010901
Price (USD)
-
Request A Quote
Type:
Spectral Library
Ask our AI about this Product

Analyze of a wide variety of common coating materials with the Thermo Scientific™ Coatings Technology Library and associated handbook.

Take the guesswork out of your spectral analysis and create greater efficiencies in your lab. Our comprehensive libraries feature fully-reviewed, high-quality spectroscopy databases, enabling confident compound identification for a variety of environmental, forensic, pharmaceutical and industrial applications.

The Coatings Technology Library includes spectra of polymers, resins, vehicles, pigments, extenders, acrylic monomers, radiation-curable monomers, fungicides, mildewcides, inhibitors, stabilizers, solvents and other additives. Samples were prepared as capillary films, melts on KBr, cast films on KRS-5 or KBr pellets.
Contains 2,507 spectra of common coating materials
  • Prepared by the Federation of Societies for Coatings Technology
  • Data format: High Resolution (HR)
About Spectral Libraries:
  • Spectral library searching has long been an extraordinarily valuable tool in the analytical chemist's toolbox. Our extensive series of spectral libraries includes information specific to application, industry or spectral technique.
  • Spectral library searching based on digital matching of spectral signatures is a very effective way of performing qualitative identification of compounds. Because FT-IR and Raman spectroscopic signatures are based on molecular functional groups, even if the exact compound is not present in the libraries, the list of matching compounds from a spectral library search can often lead to identification—or at least classification—of the material.
  • Thermo Scientific libraries span applications for Hazmat, Forensics, Food, Polymers and Organic Chemistry.
Note:

Members of the Federation of Societies for Coatings Technology qualify for a 10% discount when purchasing this library. Please be sure to provide your member number when ordering.

Includes:

Revised edition of An Infrared Spectroscopy Atlas for the Coatings Industry (1991). The atlas is the fourth edition of this FSCT publication, dealing with the use of FT-IR in coatings analysis—a 2-volume hardcover book set with valuable reference information on spectroscopy theory, analytical instrumentation, sampling accessories, sample preparation, spectral interpretation and quantitative analysis.
Specifications
For Use With (Application)FT-IR
TypeSpectral Library
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link