Thermo Scientific™

Aldrich Collection of FT-IR Spectra Edition II

Catalog number: 834-016201
Have Questions?
Thermo Scientific™

Aldrich Collection of FT-IR Spectra Edition II

Catalog number: 834-016201
Have Questions?
Effectively perform qualitative identification of compounds with the Thermo Scientific™ Aldrich™ Collection of FT-IR Spectra Edition II, comprehensive collection of FT-IR spectral references.

Take the guesswork out of your spectral analysis and create greater efficiencies in your lab. Our comprehensive libraries feature fully-reviewed, high-quality spectroscopy databases, enabling confident compound identification for a variety of environmental, forensic, pharmaceutical and industrial applications.
 
Catalog Number
834-016201
Unit Size
Each
Description
Aldrich Collection of FT-IR Spectra Edition II
Price (USD)
Full specifications
DescriptionAldrich Collection of FT-IR Spectra Edition II
For Use With (Application)Spectral Library
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
834-016201Full specifications
EachAldrich Collection of FT-IR Spectra Edition IIRequest A Quote
DescriptionAldrich Collection of FT-IR Spectra Edition II
For Use With (Application)Spectral Library
Unit SizeEach
Showing 1 of 1
This high resolution library contains the most common chemicals found in The Aldrich Handbook of Fine Chemicals
  • Contains spectra of 18,454 pure compounds
  • Data format: High Resolution (HR)
  • All spectra were acquired by Sigma-Aldrich, and were processed by Thermo Fisher Scientific
  • Represents the largest Aldrich collection of spectra collected entirely by FT-IR instrumentation
  • Eight smaller Aldrich Material Specific Sub-Libraries are also available
  • Includes The Second Edition of the Aldrich Library of FT-IR Spectra (a three-volume book set)
About Spectral Libraries:
  • Spectral library searching has long been an extraordinarily valuable tool in the analytical chemist's toolbox. Our extensive series of spectral libraries includes information specific to application, industry or spectral technique.
  • Spectral library searching based on digital matching of spectral signatures is a very effective way of performing qualitative identification of compounds. Because FT-IR and Raman spectroscopic signatures are based on molecular functional groups, even if the exact compound is not present in the libraries, the list of matching compounds from a spectral library search can often lead to identification—or at least classification—of the material.
  • Thermo Scientific libraries span applications for Hazmat, Forensics, Food, Polymers and Organic Chemistry.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text