Nicolet™ Condensed Phase Sampler FTIR Spectral Library
Nicolet™ Condensed Phase Sampler FTIR Spectral Library
Thermo Scientific™
Nicolet™ Condensed Phase Sampler FTIR Spectral Library
Facilitate high productivity for investigative laboratories with this cost-effective, general purpose library, a collection of infrared spectra from multiple libraries.
Have Questions?
Catalog number 834-018300
Price (USD)/ Each
-
Request A Quote
Description:
Nicolet Condensed Phase Sampler FTIR Spectral Library
Facilitate high productivity for investigative laboratories with this cost-effective, general purpose library, a collection of infrared spectra from multiple libraries. The Thermo Scientific™ Nicolet™ Condensed Phase Sampler FTIR Spectral Library is designed to cover a wide range of investigative needs.
Take the guesswork out of your spectral analysis and create greater efficiencies in your lab. Our comprehensive libraries feature fully-reviewed, high-quality spectroscopy databases, enabling confident compound identification for a variety of environmental, forensic, pharmaceutical and industrial applications.
Consists of 842 spectra of drugs, polymers, inorganics, and general chemicals that can be used for investigation of unknown samples
  • Data format: High Resolution (HR)
About Spectral Libraries
Spectral library searching has long been an extraordinarily valuable tool in the analytical chemist's toolbox. Our extensive series of spectral libraries includes information specific to application, industry or spectral technique.
Spectral library searching based on digital matching of spectral signatures is a very effective way of performing qualitative identification of compounds. Because FT-IR and Raman spectroscopic signatures are based on molecular functional groups, even if the exact compound is not present in the libraries, the list of matching compounds from a spectral library search can often lead to identification, or at least classification, of the material.
Thermo Scientific libraries span applications for Hazmat, Forensics, Food, Polymers and Organic Chemistry.
Specifications
DescriptionNicolet Condensed Phase Sampler FTIR Spectral Library
For Use With (Application)FT-IR
Unit SizeEach