Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Streamline your routine, complex and advanced routines and workflows with Thermo Scientific™ Insight™ Pro Software. Prevent costly sampling and measurement errors with prompting, step-by-step workflows. Add Thermo Scientific™ Insight™ Pro Security Software to achieve compliance with current FDA 21 CFR Part 11 regulations.
When installation and operational qualifications (IQ/OQ) are required, Thermo Scientific™ Validator™ for Insight Pro Software provides all the documentation and reference materials needed to complete IQ/OQ for Thermo Scientific™ Evolution™ UV-Visible Spectrophotometers.
Certifications/Compliance | Supports current 21 CFR Part 11 compliance |
Type | Insight Pro Security Software |
For Use With (Equipment) | For use with Thermo Scientific Evolution One, One Plus, and Pro UV-Vis Spectrophotometers |
Unit Size | Each |
Type | Insight Pro Software |
For Use With (Equipment) | For use with Thermo Scientific Evolution One, One Plus, and Pro UV-Vis Spectrophotometers |
Unit Size | Each |
Description | Features: IQ/OQ procedures for spectrophotometers, accessories, and software, Easy to follow steps that reference the included uncertified filter set, High quality binder for 8.5 inch x 11 inch paper, Integrated sign-off in the IQ streamlines the process, Test result sheets for OQ procedures saves paper and makes qualification easyIncludes: Binder with pre-printed tabs and DVD with: IQ/OQ procedures for spectrophotometers, accessories, and software, Concepts of validation documentation, Design Qualification section detailing vendor practices, Best practice guide for Performance Verification, Preventive maintenance information and forms, Incident report sheets and template masters, Preprinted master copies of certain core documents are included, Documents not required by all users must be printed from the DVD, Spectrophotometer Test Kit with calibrated filters (840-285200) referenced in the qualification procedures. |
Type | Validator for Insight Pro - US Version |
For Use With (Equipment) | For use with Thermo Scientific Evolution One, One Plus, and Pro UV-Vis Spectrophotometers |
Unit Size | Each |
Description | Features: IQ/OQ procedures for spectrophotometers, accessories, and software, Easy to follow steps that reference the included uncertified filter set, High quality binder A4 paper, Integrated sign-off in the IQ streamlines the process, Test result sheets for OQ procedures saves paper and makes qualification easy Includes: Binder with pre-printed tabs and DVD with: IQ/OQ procedures for spectrophotometers, accessories, and software, Concepts of validation documentation, Design Qualification section detailing vendor practices, Best practice guide for Performance Verification, Preventive maintenance information and forms, Incident report sheets and template masters, Preprinted master copies of certain core documents are included, Documents not required by all users must be printed from the DVD, Spectrophotometer Test Kit with calibrated filters (840-285200) referenced in the qualification procedures. |
Type | Validator for Insight Pro - A4 Version |
For Use With (Equipment) | For use with Thermo Scientific Evolution One, One Plus, and Pro UV-Vis Spectrophotometers |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Type | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
837-004000 | Each | Insight Pro Security Software | 4,470.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
837-003900 | Each | Insight Pro Software | 1,050.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
840-370900 | Each | Validator for Insight Pro - US Version | 2,930.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
840-371000 | Each | Validator for Insight Pro - A4 Version | Request A Quote | - | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Make your lab time more productive with Insight Pro Software
Streamlined QA/QC Analysis:
Simplified Research Applications:
Integrated Accessories:
Comply with US FDA 21 CFR Part 11:
To ensure the security and integrity of your data and to help your lab comply with 21 CFR Part 11 or other regulations, add Insight Pro Security Software with which pairs Insight Pro Software with Thermo Scientific™ Security Suite Software.
Instrument Qualification:
Validator for Insight Pro Software includes: