Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DILYVKDLAF VDPDDCTPLK TITRFYNHPV HFVFHDTKLD AMLEEFKKGK SHLAIVQKVN NEGEGDPFYE VLGLVTLEDV IEEIIKSEIL DESDMYTDNR SRKRVSEKNK RDFSAFKDAD NELKVKISPQ LLLAAHRFLA TEVSQFSPSL ISEKILLRLL KYPDVIQELK FDEHNKYYAR HYLYTRNKPA DYFILILQGK VEVEAGKENM KFETGAFSYY GTMALTSVPS DRSPAHPTPL SRSASLSYPD RTDVSTAATL AGSSNQFGSS VLGQYISDFS VRALVDLQYI KITRQQYQNG LLASRMENSP QFPIDGCTTH MENLAEKSEL PVVDETTTLL NERNSLLHKA SHENAI
This gene encodes a member of the ancient conserved domain containing protein family. Members of this protein family contain a cyclin box motif and have structural similarity to the cyclins. The encoded protein may play a role in metal ion transport. Mutations in this gene are associated with Jalili syndrome which consists of cone-rod dystrophy and amelogenesis imperfecta.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ancient conserved domain protein 4; Ancient conserved domain-containing protein 4; cyclin M4; Cyclin-M4; Metal transporter CNNM4
Gene Aliases: ACDP4; CNNM4; KIAA1592
UniProt ID: (Human) Q53RE5
Entrez Gene ID: (Human) 26504
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support