Holmium and Didymium Glass Standards
Product Image
Thermo Scientific™
Holmium and Didymium Glass Standards
Thermo Scientific™ Wavelength Calibration Filter Sets consist of Holmium and Didymium solid filters traceable to international standards.
Have Questions?
Catalog NumberFor Use With (Equipment)
840-253200Standard 10mm pathlength square cuvette holder in any spectrophotometer
Catalog number 840-253200
Price (USD)
-
Request A Quote
For Use With (Equipment):
Standard 10mm pathlength square cuvette holder in any spectrophotometer
Thermo Scientific™ Wavelength Calibration Filter Sets consist of Holmium and Didymium solid filters traceable to international standards.
  • Calibrated according to a certified process in our NVLAP accredited calibration laboratory in Madison, WI, USA.  (NVLAP Code:  200955-0)
  • Mounted in a filter holder suitable for use with any UV-Visible spectrophotometer sample holder
  • Calibrated at eight peak wavelengths for Holmium and five for Didymium
Specifications
Certifications/ComplianceTraceable to mercury emission lines, a recognized fundamental standard
DescriptionHolmium Didymium Glass Standards
For Use With (Equipment)Standard 10mm pathlength square cuvette holder in any spectrophotometer
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link