1x1 image pixel for data collection
NanoDrop™ One/OneC Security Suite Compliance Software
NanoDrop™ One/One<sup>C</sup> Security Suite Compliance Software
Thermo Scientific™
NanoDrop™ One/OneC Security Suite Compliance Software
Ensure downstream application success with a microvolume spectrophotometer that accurately measures concentration and purity from just 1-2 μL of nucleic acid or protein samples.
Have Questions?
Catalog NumberType
840-329700Security Suite for ND One/OneC Spectrophotometer
Catalog number 840-329700
Price (USD)
2,815.00
Each
Add to cart
Type:
Security Suite for ND One/OneC Spectrophotometer
Price (USD)
2,815.00
Each
Add to cart
Confidently secure your laboratory data and more easily achieve compliance with US FDA 21 CFR Part 11 with Thermo Scientific™ Security Suite Software for use with the NanoDrop™ One/ OneC Microvolume Spectrophotometer. The software helps to facilitate compliant results in regulated environments with advanced tools such as user account access control, electronic signatures, audit trails, automated data exports, and security policies. Security Suite software integrates directly into the NanoDrop One PC Control Software for streamlined operation.
Security Suite for NanoDrop One/OneC
Software is designed to integrate into the free NanoDrop One/OneC PC control software.
Suite includes:
  • Thermo Scientific Security Administration and Thermo Scientific Audit Manager
    These two pieces of software enable users to run the NanoDropOne/OneC PC control software in a lab that needs to comply with FDA 21 CFR Part 11.
Software features include:
  • User account access controlled through Windows log-in
  • Electronic signaturesAudit trail
  • Automated data export
  • Security policies (example: prevent data deletion)
Included with the order:
  • USB flash drive containing the Security Suite for the NanoDrop One/OneC PC control software installer
Specifications
Certifications/Compliance21 CFR Part 11 compliance
For Use With (Equipment)NanoDrop One/OneC Spectrophotometers
IncludesThermo Scientific Security Suite Software
DisplayDigital
TypeSecurity Suite for ND One/OneC Spectrophotometer
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link