Evolution™ One/One Plus UV-Vis Spectrophotometers
Evolution™ One/One Plus UV-Vis Spectrophotometers
Evolution™ One/One Plus UV-Vis Spectrophotometers
Thermo Scientific™

Evolution™ One/One Plus UV-Vis Spectrophotometers

Green features
Thermo Scientific™ Evolution™ UV-Vis Spectrophotometers combine reliable and versatile hardware with easy-to-learn, easy-to-use Thermo Scientific™ Insight™ Pro Software. Together they will meet the diverse needs your organization requires from an analytical tool.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeSpectral Bandwidth
840-341400Evolution One UV-Vis Spec1 nm
840-341500Evolution One Plus UV-Vis Spec1.0, 2.0 nm Variable
840-370100Evolution One US Security Bundle1 nm
840-370500Evolution One INTL Security Bundle1 nm
840-370200Evolution One US USP/EP Standards Bundle1 nm
840-370600Evolution One INTL USP/EP Standards Bundle1 nm
840-370400Evolution OnePlus US Security Bundle1.0, 2.0 nm Variable
840-370800Evolution OnePlus INTL Security Bundle1.0, 2.0 nm Variable
840-370300Evolution OnePlus US USP/EP Standards Bundle1.0, 2.0 nm Variable
840-370700Evolution OnePlus INTL USP/EP Standards Bundle1.0, 2.0 nm Variable
Catalog number 840-341400
Price (USD)
10,200.00
Each
Type:
Evolution One UV-Vis Spec
Spectral Bandwidth:
1 nm
Price (USD)
10,200.00
Each

For routine, complex research and advanced applications, including current compliance, the Thermo Scientific™ Evolution™ One/One Plus UV-Vis Spectrophotometer series brings together reliable, versatile hardware, a broad range of accessories, with easy-to-use, easy-to-learn Thermo Scientific™ Insight™ Pro Software.

Webinar: Identification of protein aggregation through UV-Vis spectroscopy

For routine to research analysis, this instrument series offers real innovations that provide enhanced usability and performance - without added complexity. With straightforward software, cutting-edge instrumentation and a wide selection of accessories, the Evolution One and One Plus UV-Vis Spectrophometers work with you to perform experiments the way you want - for the results you need.

Optimized Engineering Delivers Ultimate Performance:
  • Double-beam geometry with fixed 1nm or variable spectral bandwidth options
  • Application Focused Beam Geometry (AFBG) technology enables the best performance from microcells, solid samples, and fiber optic probes (Evolution One Plus Only)
  • Xenon flash lamp requires no warm-up time allowing instant measurements
  • Meet chemical, materials science, or biological sampling and measurement needs with a broad range of accessories
Insight Pro Software—Easy-to-learn, easy-to-use
  • Reduce training time: Optimized user interface with prompting software defines clear step-by-step workflow and is easy to teach and learn
  • Prevent costly sampling and measurement errors: Create dedicated workflows for complex assays and transform multi-step workflows into one simplified, easy-to-follow method suitable for any level of user with Customized User Environment (CUE)
  • Always current: Whether it is user inputs or application requirements, or new regulations, Insight Pro Software is continuously improving with quick upgrades to deliver greater capabilities and efficiencies for your lab’s needs
Accessories for all your Sampling Needs:
  • Wide accessory selection with various temperature control options, sample changers, sippers and solid sampling devices complete your analysis
  • Choice of Peltier Single Cell Holder or a Smart 8-Cell Peltier system for temperature control and sample monitoring from 0° to 100 °C
  • Easy software integration gives you complete control of your measurements

Regulatory/Quality Control Environments:
To use the EvolutionOne/One Plus UV-Vis Spectrophotometers in a regulated or quality control environment, we offer a wide selection of compliance products and services to reduce implementation time and assure system performance.

Evolution One/One Plus Security and Validator Bundles include:
  • Evolution One or One Plus UV-Vis Spectrometer
  • INSIGHT Pro with Security Software
  • Non-USP and EP Standards Set
Evolution One/One Plus USP/EP Standards Pharma Bundles include:
  • Evolution One or One Plus Spectrometer
  • INSIGHT Pro with Security Software
  • USP and EP UV Standards Set
  • USP and EP Visible Standards Set
Specifications
Baseline Flatness±0.001 A, 200–800 nm, 1.0 nm SBW, smoothed
Dimensions (L x W x H)593 x 475 x 266 mm (23.3 x 18.7 x 10.6 in.)
Drift<0.0005 A/hr., 500 nm, 1.0 nm SBW, 1 hour warmup
Electrical Requirements100/240 V, 50/60 Hz selected automatically, 150 W maximum
Green FeaturesACT Ecolabel (Gold)
IncludesEvolution One Plus UV-Vis Spectrometer
Lamp Life>5 years or longer if not using live signal
Noise
0A: ≤0.00015 A
1A: ≤0.00025 A
2A: ≤0.00050 A
260 nm, 1.0 nm SBW, RMS<
Optical DesignDouble-beam with sample and reference cuvette positions Czerny-Turner Monochromator
Pharmacopoeia Compliance TestingPhotometric Accuracy (60mg/L K2Cr2O7): ±0.010A
Stray Light: ≤1%T at 198 nm: KCI; ≤0.05%AT at 220 nm: Nal, Kl
Wavelength Accuracy: ±.5 nm 541.9, 546.1nm Hg emission lines, ±0.8 nm full range
Wavelength Repeatability: ≤0.05 nm, repetitive scanning of 546.1 nm Hg emission line
Photometric Accuracy1 A: ±0.004 A, 2 A: ±0.008 A, Measured at 440 nm using neutral density filters traceable to NIST
Photometric Display-0.3 to 4.0 A
Photometric Range>3.5 A
Photometric Repeatability1 A: ±0.0002 A
Scan Ordinate ModesAbsorbance, % Transmittance, % Reflectance, Kubelka-Munk, log (1/R), log (Abs), Abs*Factor, Intensity
Scan Speed<1 to 6000 nm/min. (Variable)
WarrantyInstrument (2 years), Lamp (3 years)
Wattage150 W max.
Wavelength Accuracy±0.5 nm (541.9, 546.1 nm mercury lines), ±0.8 nm (full range 190 to 1100 nm)
Wavelength Data Interval10, 5, 2, 1.0, 0.5, 0.2, 0.1 nm
Wavelength Repeatability≤0.05 nm (546.1 nm mercury line, SD of 10 measurements)
Weight (English)32 lb.
Weight (Metric)14.5 kg
Detector TypeDual Silicon Photodiodes
Light SourceXenon Flash
Product LineEvolution One
Spectral Bandwidth1 nm
TypeEvolution One UV-Vis Spec
Wavelength Range190 to 1100 nm
Unit SizeEach