Nebulizers for ICP-OES Analyzers
Nebulizers for ICP-OES Analyzers
Actual product may vary
Thermo Scientific™
Nebulizers for ICP-OES Analyzers
Enhance your analysis capabilities with a range of nebulizers compatible with the Thermo Scientific™ ICP-OES analyzers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
842312051331AeroSalt High Solids Nebulizer
842312051321V Groove Nebulizer
842312051471Burgener Mira Mist Nebulizer
842312051431Standard Nebulizer
Catalog number 842312051331
Price (USD)
468.00
Each
Add to cart
Type:
AeroSalt High Solids Nebulizer
Price (USD)
468.00
Each
Add to cart
Enhance your analysis capabilities with a range of nebulizers compatible with the Thermo Scientific™ iCAP™ Series ICP-OES systems. Choose one to suit your sample matrix to ensure optimum performance.
Enhanced Performance through Advanced Accessories
These Nebulizers are available as standalone items, or as part of the Thermo Scientific™ ICP-OES Sample Introduction Kits.
  • Standard Nebulizer handles nonaggressive liquid samples
  • High Solids Nebulizer is intended for the analysis of samples containing relatively high solids (up to 15%)
  • Burgener MiraMist Nebulizer, PEEK components handle most liquids, except for a few acids and organic solvents
  • V Groove Nebulizer is intended for the analysis of samples containing very high solids (30% or more) and for analysis of nonvolatile organic solvents
Specifications
For Use With (Equipment)iCAP Series ICP-OES Systems
TypeAeroSalt High Solids Nebulizer
Unit SizeEach
Share catalog number, name or link