1x1 image pixel for data collection
Argon Humidifier Tubing Kit
Argon Humidifier Tubing Kit
Thermo Scientific™
Argon Humidifier Tubing Kit
This convenient Thermo Scientific™ Argon Humidifier Tubing Kit contains all the tubing you need to run ICP-OES analysis with our Argon Humidifier accessory.
Have Questions?
Catalog NumberType
842312052101Argon Humidifier Tubing Kit
Catalog number 842312052101
Price (USD)
-
Request A Quote
Type:
Argon Humidifier Tubing Kit
Ask our AI about this Product
For ultimate convenience, use this Thermo Scientific™ Argon Humidifier Tubing Kit, containing all the consumable tubing you need to run ICP-OES analysis with our Argon Humidifier accessory, for the analysis of high dissolved solids samples, including soils.
Specifications
For Use With (Equipment)ICP-OES Analyzers
TypeArgon Humidifier Tubing Kit
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection