Sprint Valve Kits for the iCAP™ 7600 ICP-OES System
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Thermo Scientific™
Sprint Valve Kits for the iCAP™ 7600 ICP-OES System
Convenient Thermo ScientificTM Sprint Valve Kits contain all the consumables required to set up Sprint analysis for ICP-OES.
Have Questions?
Change view
Catalog Number
Type
842312052641
Organics Sprint Valve Kit
842312052631
Aqueous Sprint Valve Kit
2 Options
Catalog number 842312052641
Price (EUR)
4 800,00
Each
Add to cart
Type:
Organics Sprint Valve Kit
Price (EUR)
4 800,00
Each
Add to cart
Ask our AI about this Product
Enable extremely fast analysis times, utilizing the integrated Sprint Valve kits for the Thermo Scientific™ iCAP™ 7600 ICP-OES system, in conjunction with the Sprint analysis mode. These two, convenient Sprint Valve Kits contain all the consumables required to set up Sprint analysis for either aqueous or organic sample matrices.
Select Customized Consumables Kits for Easier Analysis
Aqueous sprint valve kit
Organics sprint valve kit
Specifications
For Use With (Equipment)iCAP 7600 ICP-OES System
TypeOrganics Sprint Valve Kit
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our
Privacy Notice.