Torch Holder Assembly Spares for the EMT Plasma Torch for the iCAP™ 7000 Series ICP-OES Systems
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Thermo Scientific™
Torch Holder Assembly Spares for the EMT Plasma Torch for the iCAP™ 7000 Series ICP-OES Systems
For optimum ICP-OES instrument performance, use only dedicated Thermo Scientific™ consumables, including spares for the EMT plasma Torch Holder Assembly.
Have Questions?
Catalog Number
Type
842315550171
Torch Holder Assembly Spares
Catalog number 842315550171
Price (EUR)
282,00
Each
Add to cart
Type:
Torch Holder Assembly Spares
Recurring order eligible. Learn more »
Price (EUR)
282,00
Each
Add to cart
Ask our AI about this Product
Keep your ICP-OES analyzer in top condition; use only dedicated spares and Thermo Scientific™ Consumables, including Thermo Scientific™ Torch Holder Assembly Spares for the EMT plasma torch for Thermo Scientific™ iCAP™ 7000 Series ICP-OES systems. Simple routine maintenance is enabled through the unique design of the Thermo Scientific™ Enhanced Matrix Tolerance (EMT) plasma torch, just clip the torch in and out in seconds.
Specifications
For Use With (Equipment)iCAP 7000 Series ICP-OES Systems
TypeTorch Holder Assembly Spares
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our
Privacy Notice.