DyLight™ Antibody Labeling Kits
Thermo Scientific™

DyLight™ Antibody Labeling Kits

Covalently label antibodies or proteins with DyLight dyes for use in immunofluorescence, flow cytometry, western blot analysis, and high content analysis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberExcitation/EmissionLabel or Dye
84530562/576DyLight 550
53020400/420DyLight 405
53024490/525DyLight 488
53044590/617DyLight 594
53046638/658DyLight 633
53056679/702DyLight 680
53062784/814DyLight 800
62275346/442DyLight 350
84535652/672DyLight 650
84538749/775DyLight 755
Catalog number 84530
Price (USD)
652.00
Each
Excitation/Emission:
562/576
Label or Dye:
DyLight 550
Price (USD)
652.00
Each
Invitrogen Dylight Antibody Labeling kits provide everything needed to label 1 mg of antibody or protein with amine-reactive Dylight dye and subsequent free dye removal. Choose from a variety of Dylight fluorophores across the spectrum to covalently label your antibody or protein. Labeled antibodies are typically ready to use in 90 minutes with minimal hands-on time. The kit contains all necessary components to perform three separate labeling reactions using 1 mg of IgG or similar quantities of other proteins.
Invitrogen DyLight Antibody Labeling kits enable the creation of fluorescent antibodies that are covalently linked to different DyLight fluorophores. The fluorescently labeled antibodies can be used in a wide variety of laboratory applications, including immunofluorescence microscopy, IHC, western blotting, ELISA, flow cytometry, or in vitro and in vivo fluorescent detection strategies.

DyLight Antibody Labeling kits contain an NHS ester-activated derivative of the high-performance DyLight dye that can be used to fluorescently label antibodies and other proteins for cellular imaging and other fluorescence detection methods. The DyLight dye is an amine-reactive dye and is activated with an N-hydroxysuccinimide (NHS) ester moiety to react with exposed N-terminal α-amino groups or the ε-amino groups of lysine residues to form stable amide bonds.

DyLight Antibody Labeling kits contain all necessary components to perform three separate labeling reactions and free dye removal using 1 mg of IgG or similar quantities of other proteins.

DyLight dyes exhibit higher fluorescence intensity and photostability than CyDye and LI-COR™ dyes in many applications and remain highly fluorescent over a broad pH range (pH 4–9). Additionally, the water solubility of the DyLight fluorescent dyes allows a high dye-to-protein ratio to be achieved without causing precipitation of conjugates. Finally, the high solubility of these fluorophores permits protein solutions to be added directly to specific amounts of the labeling reagent.

Other advantages of the DyLight Antibody Labeling kits:
· High performance—high fluorescence intensity across a broad pH range
· Multiple colors—10 different dyes across the visible spectrum and extending into the infrared
· Specificity—NHS ester-activated dye labels proteins and other molecules at primary amines (-NH2)
· Optimized procedure—following the standard protocol results in antibodies with excellent dye:protein ratios and recovery rates for optimum activity and fluorescence labeling

We also offer DyLight Microscale Antibody Labeling kits for fast and efficient fluorescent labeling of antibodies for use in fluorescence methods. The DyLight Microscale Antibody Labeling kits contain all necessary components to perform five separate labeling reactions using 100 μg of IgG or similar quantities of other proteins. Both the standard and microscale kit sizes include the amine-reactive DyLight fluorophore in convenient single-use vials as well as purification resin and spin columns for the preparation of ready-to-use conjugate.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorYellow
Excitation/Emission562/576
Kit ContentsContains 3 vials DyLight NHS Ester, 1 mL Tube Borate Buffer, 5 mL Purification Resin, 6 Spin Columns, and 12 Microcentrifuge Collection Tubes
Label TypeDyLight™
Labeling Scale0.5 to 1 mg
Product LineDyLight
Product TypeLabeling Kit
Quantity3 reactions
Chemical ReactivityAmine
Labeling TargetAntibodies, Proteins
Label or DyeDyLight 550
Unit SizeEach
Contents & Storage
Store the DyLight NHS Ester at -20°C. Store all other kit components at 4°C.