Thermo Scientific™
Western Blot Roller
Catalog number: 84747
Have Questions?
Thermo Scientific™
Western Blot Roller
Catalog number: 84747
Have Questions?
Catalog Number
84747
Unit Size
1 unit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
847471 unit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific Western Blot Roller is a simple, reusable tool for gently and easily removing trapped air bubbles when assembling transfer stacks in semi-dry or wet-tank transfer cassettes.
The transfer-stack roller has a stainless steel handle and an inert PTFE roller that is 8.4 cm wide. Mini-gel sandwiches can be rolled out with one or two passes, but the tool also works well for transfer stacks with larger gels. The tool is included as an accessory with the Pierce Power Blotter (Part No. 22834).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)
Pierce™ Power System
Mode of Transfer
Semi-Dry, Wet
Type
Western Blot Roller
Contents & Storage
A 8.4 cm-wide PTFE roller with stainless steel handle