Triton™ X-100 Surfact-Amps™ Detergent Solution
Triton™ X-100 Surfact-Amps™ Detergent Solution
Thermo Scientific™

Triton™ X-100 Surfact-Amps™ Detergent Solution

Thermo Scientific Triton X-100 Surfact-Amps Detergent Solution is a highly-purified Triton™ X-100 detergent stabilized as a 10% solution (w/v) inRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
8511150 mL
283146 x 10 mL
85112250 mL
283131 L
Catalog number 85111
Price (USD)
129.65
Online Exclusive
147.00
Save 17.35 (12%)
Each
-
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (USD)
129.65
Online Exclusive
147.00
Save 17.35 (12%)
Each
Add to cart
Ask our AI about this Product
Thermo Scientific Triton X-100 Surfact-Amps Detergent Solution is a highly-purified Triton™ X-100 detergent stabilized as a 10% solution (w/v) in variable sizes packaged under nitrogen in 10 mL glass ampules or three sizes of non-leaching HDPE bottles, ensuring their stability and eliminating the accumulation of peroxides and degradation products.

Features of Triton X-100 Surfact-Amps Detergent Solution:

Triton X-100—a nonionic detergent for use in various protein methods
Accurate—precise 10% detergent solution in ultrapure water
Easy-to-use—solution is simple to dispense and dilute for use
Exceptionally pure—less than 1.0μeq/mL peroxides and carbonyls
Stable—packaged under inert nitrogen gas in glass ampules or HDPE bottles

Specifications for Triton X-100 Surfact-Amps Detergent Solution:
• Visual: Clear to slightly hazy, colorless solution, free of particulate matter.
• Concentration: 10.0±1.0%
• Oxidants: ≤1.0μeq/mL
• Carbonyls: ≤1.0μeq/mL
• Suspended Solids: Residue present must not exceed Residue Reference.

Properties of Triton X-100:
• Molecular Weight: 647g
• Detergent Class: Nonionic polyoxyethylene surfactant
• Aggregation Number: 140
• Micelle Molecular Weight: 90,000g
• Critical Micelle Concentration (CMC): 0.24 mM (0.0155% w/v)
• Cloud Point: 64°C
• Dialyzable: No

Triton X-100 is most frequently used as a component of cell lysis buffers or other solutions intended to extract and solubilize proteins.

Related Products
Surfact-Amps™ Detergent Sampler
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity50 mL
Reagent TypeDetergent Solution
FormLiquid
Product LineSurfact-Amps™, Triton™
Product TypeDetergent
Unit SizeEach
Contents & Storage
Store between 20°C and 25°C
Learn about immunopeptidomics basics
Whether you're new to or experienced with proteomics, we're here to help.


Learn about immunopeptidomics basics ›
Follow 5-step sample prep workflow ›

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

I need to do immunolabeling of a thick cryosection, around 60 µm thick. What should I do to ensure good labeling with my antibody?

One thing you should do is to permeabilize well, by including 0.1% to 0.2% Triton X-100 in all washes and incubations. Another recommendation is to extend the labeling times to overnight at 4oC for both primary antibody and secondary antibody. Finally, label the section free-floating, rather than adhered to a slide, to gain penetration for the antibody from both sides.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What are the storage conditions for Triton X-100 Surfact-Amps Detergent Solution (Cat. No. 85111)?

After opening, we recommend storing the unused solution sealed in an air-tight vial containing minimal air space or from which the air has been purged with nitrogen gas. Frequent opening should be avoided because Triton X-100 is susceptible to autoxidation upon exposure to air.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can you provide the shelf life for Triton X-100 Surfact-Amps Detergent Solution (Cat. No. 85111)?

Triton X-100 Surfact-Amps Detergent Solution is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (20-25 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What are the main advantages of using Thermo Scientific Pierce Surfact-Amps detergents?

Thermo Scientific Pierce Surfact-Amps detergents are highly purified, precisely diluted (10%) formulations that are ideal for applications or assays that are sensitive to contaminants that are present in unpurified detergents. We test every batch to insure that our detergents contain less than 1.0 µeq/mL peroxides and carbonyls and package them under nitrogen, to prevent oxidization during storage.

Superior quality - lower measurable contaminant levels than other leading vendors
Accurate -precise 10% detergent solution in ultrapure water
Easy-to-use - solution is simple to dispense and dilute for use
Exceptionally pure - less than 1.0 µeq/mL peroxides and carbonyls

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Are detergents denaturing or non-denaturing with respect to protein structure?

Ionic detergents, or those that carry a charge, are the most likely to be denaturing to proteins. Denaturing detergents can be anionic such as sodium dodecyl sulfate (SDS) or cationic such as ethyl trimethyl ammonium bromide. These detergents totally disrupt membranes and denature proteins by breaking protein-protein interactions through changes in the three-dimensional structure of the proteins. Nondenaturing detergents can be divided into nonionic detergents (i.e., Triton X-100), bile salts (i.e., cholate), and zwitterionic detergents (i.e., CHAPS).



Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link