Thermo Scientific™

Tween™ 20 Surfact-Amps™ Detergent Solution

Catalog number: 85114
Have Questions?
Thermo Scientific™

Tween™ 20 Surfact-Amps™ Detergent Solution

Catalog number: 85114
Have Questions?
Catalog Number
85114
Unit Size
250 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
6 x 10 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
85114250 mL
-
8511350 mL
-
283206 x 10 mL
-
85115500 mL
-
283211 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Tween-20 Surfact-Amps Detergent Solution is a highly-purified Tween™ 20 detergent stabilized as a 10% solution (w/v) and packaged under nitrogen in glass ampules or non-leaching HDPE bottles, ensuring it's stability and eliminating the accumulation of peroxides and degradation products.

Features of Tween-20 Surfact-Amps Detergent Solution:

Tween 20—a popular nonionic detergent for use in ELISA, Western blotting and other immunoassay PBS or TBS wash buffers
Accurate—precise 10% detergent solution in ultrapure water
Easy-to-use—solution is simple to dispense and dilute for use
Exceptionally pure—less than 1.0μeq/mL peroxides and carbonyls
Stable—packaged under inert nitrogen gas in glass ampules or HDPE bottles

Properties of Tween 20:
• Molecular Weight: 1228g
• Detergent Class: Nonionic polyoxyethylene surfactant
• Aggregation Number: Unknown
• Micelle Molecular Weight: Unknown
• Critical Micelle Concentration (CMC): 0.06 mM (0.0074%, w/v)
• Cloud Point: 95°C
• Dialyzable: No

Specifications for Tween 20 Surfact-Amps Detergent Solution:
• Visual: Clear, light yellow liquid, free of particulates.
• Concentration: 10.0±1.0%
• Oxidants: ≤1.0μeq/mL
• Carbonyls: ≤1.0μeq/mL
• Suspended Solids: Residue present must not exceed Residue Reference.

Related Products
Surfact-Amps™ Detergent Sampler
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form
Liquid
Product Line
Surfact-Amps™, Tween™
Reagent Type
Detergent Solution
Quantity
250 mL

Contents & Storage

Store in a cool, dry, and well-ventilated area

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text