1x1 image pixel for data collection
Brij™-35 Surfact-Amps™ Detergent Solution
Brij™-35 Surfact-Amps™ Detergent Solution
Thermo Scientific™

Brij™-35 Surfact-Amps™ Detergent Solution

This Thermo Scientific™ formulation of Brij™ 35 is a stabilized 10% (w/v) solution of Brij-35 detergent for use in variousRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
85118500 mL
283166 x 10 mL
8511750 mL
Catalog number 85118
Price (USD)
1,038.65
Online Exclusive
1,134.00
Save 95.35 (8%)
Each
In stock
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)
1,038.65
Online Exclusive
1,134.00
Save 95.35 (8%)
Each
Add to cart
This Thermo Scientific™ formulation of Brij™ 35 is a stabilized 10% (w/v) solution of Brij-35 detergent for use in various protein methods. Brij™ 35 is a nonionic polyoxyethylene surfactant that is most frequently used as a component of cell lysis buffers or a surfactant in various HPLC applications.

Properties of Brij 35 Detergent:

• Molecular Weight: 1225g
• Detergent Class: Nonionic
• Aggregation Number: 40
• Micelle Molecular Weight: 49,000g
• Critical Micelle Concentration (CMC): 0.09 mM (0.011%, w/v)
• Cloud Point: >100°C
• Dialyzable: No

Features of this Thermo Scientific detergent solution:

Brij 35—a nonionic detergent for use in various protein methods
Accurate—10% detergent solution in ultrapure water
Easy-to-use—solutions are simple to dispense and dilute for use

Specifications of this Thermo Scientific Brij 35, 10% Solution:

• Visual: Clear, colorless viscous liquid, free of foreign material.
• Concentration: Concentration: 27.0 to 30.0%

Related Products
Brij™-35 Surfact-Amps™ (10% w/v) Solution
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity500 mL
Reagent TypeDetergent Solution
FormLiquid
Product LineBrij™, Surfact-Amps™
Unit SizeEach
Contents & Storage
Store at Room Temperature

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection