Thermo Scientific™

Pierce™ High Capacity Streptavidin Chromatography Cartridges, 1 mL

Catalog number: 87739
Have Questions?
Thermo Scientific™

Pierce™ High Capacity Streptavidin Chromatography Cartridges, 1 mL

Catalog number: 87739
Have Questions?
Catalog Number
87739
Unit Size
2 x 1 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 x 5 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
877392 x 1 mL
-
877401 x 5 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific™ PierceTM High Capacity Streptavidin Chromatography Cartridges are convenient, ready-to-use devices for separating biotinylated molecules from non-biotinylated molecules and for purifying antigens using biotinylated antibodies. The chromatography cartridges contain a beaded resin of immobilized recombinant streptavidin protein that binds biotinylated proteins at greater than 10 mg/mL of beads. This corresponds to 2 to 3 times higher biotin-binding capacity than products from other suppliers and is ideal when a high amount of recovery is required.

These cartridges enable fast, easy, and reproducible chromatographic separations and are compatible with the major automated liquid-chromatography systems or manual syringe processing (see Table 1 for cartridge general properties). The cartridges attach directly to ÄKTATM or FPLC systems without additional connectors. The included accessory pack readily adapts cartridges for use with Luer-LokTM syringe fittings or 1/16' tubing. These cartridges can be regenerated multiple times for affinity purification.

Features of High Capacity Streptavidin Agarose:
Streptavidin—purified recombinant streptavidin protein (53 kDa, near-neutral pI); tetrameric with four biotin-binding site per molecule
Agarose resin—support is cross-linked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
Inert and stable—streptavidin is immobilized by charge-free, leach-resistant covalent bonds, resulting in low nonspecific binding and enabling multiple uses without decline in yield
High capacity—this variety of beads has a dense load of immobilized streptavidin, providing a binding capacity greater than 10 mg of biotinylated BSA per mL of resin
Two formats—available in several bottle sizes of resin slurry and as packed 1 mL and 5 mL chromatography cartridges

Properties of cross-linked 6% beaded agarose (CL-6B):
• Support pH stability: 2 to 14 (short term); 3 to 13 (long term)
• Average particle size: 45 to 165 microns
• Exclusion limit: 10,000 to 4,000,000 daltons
• Maximum recommended flow rate: 4 mL/minute (1 mL column), 5 mL/min (5 mL column)
• Maximum linear velocity: 30 cm per hour
• Maximum pressure: 0.3 MPa, 43.5 psi or 3 bar

Pierce High Capacity Streptavidin Chromatography Cartridges consist of purified recombinant streptavidin covalently immobilized at a high density onto high-quality cross-linked 6% beaded agarose. Biotinylated antibodies, proteins, peptides, nucleic acids, and other molecules or interaction complexes can be captured, immunoprecipitated, removed, or purified from samples using this streptavidin resin.

Streptavidin is a tetrameric protein containing four biotin-binding sites similar to avidin. The native protein from Streptomyces avidinii is carbohydrate-free, has an acidic pI of 5.5 and a mass of 75 kDa. The recombinant protein used to manufacture Pierce streptavidin resins has a near-neutral isoelectric point and a mass of 53 kDa. Streptavidin generally has less nonspecific binding than avidin (from chicken egg white) because of the absence of carbohydrates.

For more information see: Chromatography Cartridges for Desalting and Affinity Purification

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Capacity (Metric)
10 mg Biotinylated BSA Binding per mL Resin
Particle Size
45 to 165 μm
Product Line
Pierce™
Quantity
2 Cartridges
Protein Form
Recombinant
Target
Biotin-Tagged Molecule
Type
Chromatography Cartridge
Stationary Phase
Streptavidin
Sufficient For
Binding ≥/= 100 μg Biotin per Cartridge (≥/= 10 mg Biotinylated BSA per Cartridge)
Format
Chromatography Cartridge
Column Type
Affinity, Agarose Resin

Contents & Storage

Upon receipt store at 4°C. Do not freeze the resin. Product is shipped at ambient temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text