Detergent Removal Spin Columns & Plates
Detergent Removal Spin Columns & Plates
Thermo Scientific™

Detergent Removal Spin Columns & Plates

Rapidly eliminate high concentrations of detergents with minimal sample loss with detergent removal resins. Thermo Scientific HiPPR (High Protein and Peptide Recovery) resin is recommended for protein concentrations up to 100 μg/mL, while Thermo Scientific Pierce resin is suitable for concentrations over 100 μg/mL.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityProduct LineProduct Type
8777625 x 125 μL ColumnsPierce™Spin Column
8778010 mLPierce™Detergent Removal Resin
8777725 x 500 μL ColumnsPierce™Spin Column
877785 x 2 mL ColumnsPierce™Spin Column
877795 x 4 mL ColumnsPierce™Spin Column
883042 x 96-well PlatesPierce™Spin Plate
883055 mLHiPPR™Spin Column
8830624 x 0.1 mL ColumnsHiPPR™Spin Column
883072 x 96-well PlatesHiPPR™Spin Plate
Catalog number 87776
Price (EUR)
197,65
Online Exclusive
257,00
Save 59,35 (23%)
Each
-
Add to cart
Quantity:
25 x 125 μL Columns
Product Line:
Pierce™
Product Type:
Spin Column
Request bulk or custom format
Price (EUR)
197,65
Online Exclusive
257,00
Save 59,35 (23%)
Each
Add to cart
Ask our AI about this Product
Detergents and surfactants used to prepare protein and peptide samples can interfere with analysis by ELISA, isoelectric focusing, and mass spectrometry (MS). Removing detergents from peptide samples is especially challenging and critical for MS analysis. Even low concentrations of detergents will contaminate instruments and interfere with column binding, elution, and peptide ionization. Thermo Scientific HiPPR and Pierce detergent removal resins are ideal for rapid removal of common detergents from tryptic digests to improve the results of LC-MS/MS and MALDI-MS analysis.
Optimized—removes >95% of detergent from samples of proteins or peptides
Fast—sample processing takes less than 15 minutes
Effective—eliminates detergent-interference in downstream applications like ELISA, isoelectric focusing, and mass spectrometry

Pierce Detergent Removal Resin
Pierce Detergent Removal Resin efficiently binds to and removes high concentrations of detergents from 10 μL to 1 mL samples with minimal sample loss. These microcentrifuge columns come prepacked with various volumes of proprietary resin and are best for processing samples with proteins or peptide concentrations greater than 100 μg/mL.

HiPPR Detergent Removal Spin Column Kit
HiPPR Detergent Removal resin has been optimized for sample concentrations of 1 to 100 μg/mL and is ideal for removing commonly used detergents, including SDS, Triton™ X-100, NP-40 and CHAPS at concentrations of 0.5-1%. The resin is available in pre-filled spin columns, 96-well filter spin plates, and as loose resin for customization.

Available formats:
Resin slurries—bulk resin supplied with empty 0.8-mL spin columns
Spin columns—0.1 mL micro-centrifuge columns
96-well spin plates—pre-dispensed 96-well filter plates, compatible with centrifugation and vacuum manifold systems for manual or automated purification

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column TypeProprietary Resin, Affinity
DescriptionMicrocentrifuge, Spin Column
Purification TargetDetergent Removal
Quantity25 x 125 μL Columns
Stationary PhaseProprietary Ligand
Sufficient For10 to 25 μL Sample Each Column
Product LinePierce™
Product TypeSpin Column
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C. The resin is stored in 0.15 M NaCl, 0.05% sodium azide.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link