Thermo Scientific™

Halt™ Protease Inhibitor Cocktail, EDTA-Free (100X)

Catalog number: 87785
Have Questions?
Thermo Scientific™

Halt™ Protease Inhibitor Cocktail, EDTA-Free (100X)

Catalog number: 87785
Have Questions?
Catalog Number
87785
Unit Size
1 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
24 x 100 µL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
877851 mL
-
7842524 x 100 µL
-
784375 mL
-
7843910 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Halt Protease Inhibitor Cocktail, EDTA-free (100X) are ready-to-use concentrated stock solutions of protease inhibitors for addition to samples to prevent proteolytic degradation during cell lysis and protein extraction.

Features of Halt Protease Inhibitor Cocktail, EDTA-free (100X):

Convenient—the refrigerator-stable, 100X liquid format is more effective and easier to use than individual inhibitors or other formats; just pipette exactly the amount you need
No proprietary ingredients—fully disclosed formulation contains optimized concentrations of six broad-spectrum protease inhibitors AEBSF, aprotinin, bestatin, E-64, leupeptin and pepstatin A stabilized in high-quality dimethylsulfoxide (DMSO)
Multiple package sizes—choose among several package sizes, including single-use microtubes that keep protease inhibitors fresh and reduce the risk of reagent contamination
EDTA-free formulations—ensures compatibility with isoelectric focusing
Lysis buffer compatible—use with Thermo Scientific Pierce Cell Lysis Buffers or nearly any other commercial or homemade detergent-based lysis reagent. Add 10 μL of concentrated cocktail per 1 mL of lysis buffer to ensure complete protection of the resulting protein extract.

Applications:
• Protection of intact, active cellular proteins from degradation by endogenous proteases
• Inhibition of protease activity from lysates produced by a variety of methods including Thermo Scientific Pierce Cell Lysis Reagents, other commercially formulated cell or tissue lysis products, sonication, French press, blender homogenization or repetitive freeze/thaw cycling
• Screening of extracts for proteolytic activity
• The study of proteolysis in the regulation of cellular processes

Proteases make up a large category of enzymes, including endopeptidases and exopeptidases, which catalyze the hydrolytic breakdown of proteins into peptides or amino acids in a wide variety of cell and tissue types. Proteases are regulated and compartmentalized in living cells, but cell lysis releases and mixes them with other extracted proteins. Their activity can thus lead to the loss of a large number of valuable proteins, adversely affecting downstream applications. For this reason, it is common practice to spike cell lysis buffers or cell extracts with a cocktails of compounds known to inactivate or inhibit proteases. Halt Protease Inhibitor Cocktails effectively inhibit serine-proteases, cysteine-proteases, aspartic acid-proteases and aminopeptidases that are typically present in cellular lysate samples. Regular and EDTA-free formulations provide for inhibition of metalloproteases and compatibility with 2D electrophoresis, respectively.

Related Products
Halt™ Protease Inhibitor Single-Use Cocktail (100X)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Description
Halt Protease Inhibitor Cocktail, EDTA-Free
Product Type
Protease Inhibitor Cocktail
Form
Liquid
Product Line
Halt™
Buffer
Lysis Buffer
Reagent Type
Protease Inhibitor
Inhibitors
Protease Inhibitor
Quantity
1 mL

Contents & Storage

Upon receipt store at 4°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text