Invitrogen™

ViewRNA™ Cell Plus Cytospin Module Kit

Catalog number: 88-19002-11
Have Questions?
Invitrogen™

ViewRNA™ Cell Plus Cytospin Module Kit

Catalog number: 88-19002-11
Have Questions?
Catalog Number
88-19002-11
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
88-19002-111 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

The ViewRNA™ Cell Plus Cytospin Module Kit is for use with the ViewRNA™ Cell Plus Assay kit (Cat. No. 88-19000). These buffers can be used to perform the assay when using cytospin slides with cells.

Please refer to the ViewRNA™ Cell Plus Assay protocol on the product data sheet for instructions for use.

Reported Application

  • Microscopy, Immunocytochemistry
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Kit
For Use With (Application)
Microscopy, Immunocytochemistry
For Use With (Equipment)
ViewRNA™ Cell Plus Assay Kit (Cat. No. 88-19000)
Product Type
Cytospin Module Kit
Quantity
1 Kit
Unit Size
1 kit

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text