Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | eBioscience Annexin V Apoptosis Detection Kit APC |
Quantity | 200 Tests |
Kit Contents | • Annexin V Binding Buffer (10X), 1 bottle (100 mL) • Annexin V APC, 1 vial (1 mL) • Propidium Iodide Staining Solution, 2 vials (0.5 mL) Note: Fixable Viability Dye eFluor 450 is not recommended for use with Annexin V Apoptosis Detection Kits. |
Product Type | Apoptosis Detection Kit |
Excitation/Emission | 650, 535/660, 617 |
Shipping Condition | Wet Ice |
Conjugate | APC, Propidium Iodide |
For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
Unit Size | 200 tests |
Description | eBioscience Annexin V Apoptosis Detection Kit FITC |
Quantity | 200 Tests |
Format | Kit |
Kit Contents | • Annexin V Binding Buffer (10X), 1 bottle (100 mL) • Annexin V-FITC conjugate, 1 vial (1 mL) • Propidium Iodide Staining Solution, 2 vials (0.5 mL) Note: Fixable Viability Dye eFluor 450 is not recommended for use with Annexin V Apoptosis Detection Kits. |
Product Type | Apoptosis Detection Kit |
Excitation/Emission | 488, 535/520, 617 |
Product Line | eBioscience |
Conjugate | FITC, Propidium Iodide |
For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
Unit Size | 200 tests |
Description | Annexin V Apoptosis Detection Kit eFluor 450 |
Quantity | 50 Tests |
Format | Kit |
Kit Contents | • Annexin V Binding Buffer (10X), 1 bottle (30 mL) • Annexin V eFluor 450, 1 vial (0.25 mL) • 7-AAD Viability Staining Solution, 1 vial (0.5 mL) Note: Fixable Viability Dye eFluor 450 is not recommended for use with Annexin V Apoptosis Detection Kits. |
Product Type | Apoptosis Detection Kit |
Excitation/Emission | 405, 546/450, 647 |
Shipping Condition | Wet Ice |
Product Line | eBioscience |
Conjugate | eFluor 450, 7-AAD |
For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
Unit Size | 50 tests |
Description | eBioscience Annexin V Apoptosis Detection Kit eFluor 450 |
Quantity | 200 Tests |
Format | Kit |
Kit Contents | • Annexin V Binding Buffer (10X), 1 bottle (100 mL) • Annexin V eFluor 450, 1 vial (1 mL) • 7-AAD Viability Staining Solution, 2 vials (0.5 mL) Note: Fixable Viability Dye eFluor 450 is not recommended for use with Annexin V Apoptosis Detection Kits. |
Product Type | Apoptosis Detection Kit |
Excitation/Emission | 405, 546/450, 647 |
Shipping Condition | Wet Ice |
Conjugate | eFluor 450, 7-AAD |
For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
Unit Size | 200 tests |
Description | eBioscience Annexin V Apoptosis Detection Kit PerCP-eFluor 710 |
Quantity | 50 Tests |
Kit Contents | • Annexin V Binding Buffer (10X), 1 bottle (30 mL) • Annexin V PerCP-eFluor 710, 1 vial (0.25 mL) Note: Fixable Viability Dye eFluor 450 is not recommended for use with Annexin V Apoptosis Detection Kits. |
Product Type | Apoptosis Detection Kit |
Shipping Condition | Wet Ice |
Conjugate | PCP-eFluor 710 |
For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
Unit Size | 50 tests |
Description | eBioscience Annexin V Apoptosis Detection Kit PerCP-eFluor 710 |
Quantity | 200 Test |
Kit Contents | • Annexin V Binding Buffer (10X), 1 bottle (100 mL) • Annexin V PerCP-eFluor 710, 1 vial (1 mL) Note: Fixable Viability Dye eFluor 450 is not recommended for use with Annexin V Apoptosis Detection Kits. |
Product Type | Apoptosis Detection Kit |
Shipping Condition | Wet Ice |
Conjugate | PCP-eFluor 710 |
For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
Unit Size | 200 tests |
Description | eBioscience Annexin V-FITC Apoptosis Detection Kit |
Quantity | 100 Tests |
Kit Contents | • Binding Buffer (4X), 1 bottle (50 mL) • rh Annexin V-FITC, 1 vial (0.5 mL) • Propidium Iodide (20 μg/mL), 1 vial (1.8 mL) |
Product Type | Apoptosis Detection Kit |
Excitation/Emission | 488, 535/520, 617 |
Shipping Condition | Wet Ice |
Conjugate | FITC, Propidium Iodide |
For Use With (Equipment) | Flow Cytometer |
Unit Size | 100 tests |
Description | eBioscience Annexin V-FITC Apoptosis Detection Kit |
Quantity | 20 Tests |
Kit Contents | • Binding Buffer (4X), 1 bottle (50 mL) • rh Annexin V-FITC, 1 vial (0.5 mL) • Propidium Iodide (20 μg/mL), 1 vial (1.8 mL) |
Product Type | Apoptosis Detection Kit |
Excitation/Emission | 488, 535/520, 617 |
Shipping Condition | Wet Ice |
Conjugate | FITC, Propidium Iodide |
For Use With (Equipment) | Flow Cytometer |
Unit Size | 20 tests |
Description | eBioscience Annexin V-FITC Apoptosis Detection Kit |
Quantity | 300 Tests |
Kit Contents | Contains: • Binding Buffer (4X), 1 bottle (50 mL) • rh Annexin V-FITC, 1 vial (1.5 mL) • Propidium Iodide (20 μg/mL), 2 vials (1.8 mL) |
Product Type | Apoptosis Detection Kit |
Excitation/Emission | 488, 535/520, 617 |
Shipping Condition | Wet Ice |
Conjugate | FITC, Propidium Iodide |
For Use With (Equipment) | Flow Cytometer |
Unit Size | 300 tests |
Description | eBioscience Annexin V Apoptosis Detection Kit PE |
Quantity | 50 Tests |
Kit Contents | Contains: • Annexin V Binding Buffer (10X), 1 bottle (30 mL) • Annexin V PE, 1 vial (0.25 mL) • 7-AAD Viability Staining Solution, 1 vial (0.5 mL) Note: Fixable Viability Dye eFluor 450 is not recommended for use with Annexin V Apoptosis Detection Kits. |
Product Type | Apoptosis Detection Kit |
Shipping Condition | Wet Ice |
Conjugate | PE, 7-AAD |
For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
Unit Size | 50 tests |
Catalog Number | Specifications | Unit Size | Conjugate | Excitation/Emission | Quantity | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
88-8007-74 | 200 tests | APC, Propidium Iodide | 650, 535/660, 617 | 200 Tests | 587.65 Online price 652.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
88-8005-74 | 200 tests | FITC, Propidium Iodide | 488, 535/520, 617 | 200 Tests | 536.65 Online price 586.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
88-8006-72 | 50 tests | eFluor 450, 7-AAD | 405, 546/450, 647 | 50 Tests | 264.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
88-8006-74 | 200 tests | eFluor 450, 7-AAD | 405, 546/450, 647 | 200 Tests | 478.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
88-8008-72 | 50 tests | PCP-eFluor 710 | 50 Tests | 238.65 Online price 276.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
88-8008-74 | 200 tests | PCP-eFluor 710 | 200 Test | 504.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
BMS500FI-100 | 100 tests | FITC, Propidium Iodide | 488, 535/520, 617 | 100 Tests | 332.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
BMS500FI-20 | 20 tests | FITC, Propidium Iodide | 488, 535/520, 617 | 20 Tests | 156.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
BMS500FI-300 | 300 tests | FITC, Propidium Iodide | 488, 535/520, 617 | 300 Tests | 552.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
88-8102-72 | 50 tests | PE, 7-AAD | 50 Tests | 187.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|