1x1 image pixel for data collection
eBioscience™ DAB Advanced Chromogenic Kit
Product Image
Invitrogen™
eBioscience™ DAB Advanced Chromogenic Kit
This DAB Advanced Chromogenic Kit contains the necessary reagents to prepare a DAB substrate working solution. The enzyme peroxidase (HorseradishRead more
Have Questions?
Catalog NumberQuantity
8801-4965-7250 tests
Catalog number 8801-4965-72
Price (USD)
116.65
Online Exclusive
120.00
Save 3.35 (3%)
Each
Add to cart
Quantity:
50 tests
Price (USD)
116.65
Online Exclusive
120.00
Save 3.35 (3%)
Each
Add to cart
This DAB Advanced Chromogenic Kit contains the necessary reagents to prepare a DAB substrate working solution. The enzyme peroxidase (Horseradish Peroxidase/HRP) utilizes the DAB substrate (3, 3’-diaminobenzidine tetrahydrochloride) producing a brownish-red precipitate. These precipiates are insoluble in organic solvent. This enzyme/substrate reaction can be used on tissue sections or cells (grown on culture slides or cytospins) to visualize the localization of an HRP-conjugated antibody in immunocytochemistry and immunohistochemistry applications.
Reported Application
Microscopy, Immunohistochemical Staining, Western Blotting, Immunocytochemistry
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodColorimetric
Product TypeDAB Advanced Chromogenic Kit
Quantity50 tests
Substrate PropertiesChemical Substrate
Substrate TypeHRP (Horseradish Peroxidase) Substrate
Product LineeBioscience™
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link