MagniSort™ Mouse CD4 Naïve T cell Enrichment Kit
Product Image
Invitrogen™
MagniSort™ Mouse CD4 Naïve T cell Enrichment Kit
The MagniSort™ Mouse CD4 Naïve T cell Enrichment Kit is designed for the magnetic separation of CD4 naïve T cellsRead more
Have Questions?
Catalog NumberQuantity
8804-6824-74200 tests
Catalog number 8804-6824-74
Price (EUR)
836,00
Each
Add to cart
Quantity:
200 tests
Price (EUR)
836,00
Each
Add to cart
Ask our AI about this Product
The MagniSort™ Mouse CD4 Naïve T cell Enrichment Kit is designed for the magnetic separation of CD4 naïve T cells by negative selection. It has been optimized for the isolation of mouse CD4 naïve T cells from spleens or lymph nodes utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads. Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave CD4 naïve T cells untouched and free in solution.
The MagniSort™ Mouse CD4 Naïve T cell Enrichment Antibody Cocktail contains the following antibodies:
Anti-Mouse CD8 Biotin
Anti-Mouse CD11b Biotin
Anti-Mouse CD19 Biotin
Anti-Mouse CD24 Biotin
Anti-Mouse CD44 Biotin
Anti-Mouse CD45R (B220) Biotin
Anti-Mouse CD49b (Integrin alpha 2) Biotin
Anti-Mouse Ly-6G (Gr-1) Biotin
Anti-Mouse γδ TCR Biotin
Anti-Mouse TER-119 Biotin
Reported Application
Magnetic Cell Separation
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeT Cells (Naïve,CD4+)
No. of Tests200 tests
Quantity200 tests
Target SpeciesMouse
Product TypeCell Enrichment Kit
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link