Thermo Scientific™

L-Lysine-2HCl, 13C6, 15N2 for SILAC

Catalog number: 88209
Have Questions?
Thermo Scientific™

L-Lysine-2HCl, 13C6, 15N2 for SILAC

Catalog number: 88209
Have Questions?
Catalog Number
88209
Unit Size
50 mg
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
8820950 mg
-
88432500 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Heavy and Light Amino Acids are used to specifically analyze protein expression by mass spectrometry using stable isotope labeling with amino acids in cell culture (SILAC) quantification kits. This is a lyophilized preparation of L-Lysine-2HCl (13C6, 15N2), sufficient for 1 SILAC experiment.

General features of heavy and light amino acids for SILAC labeling:

Efficient—100% label incorporation into proteins of living cells
Flexible—different isotopes of heavy and/or light amino acids for arginine, lysine, leucine and proline enable the quantitation of peptides derived from MS-grade proteases
Multiplex capabilities—several alternative isotopes of arginine and lysine are available that allow the analysis of multiple treatment conditions in each experiment
High-quality supplements—heavy amino acids with >99% isotope purity

Stable isotope labeling with amino acids in cell culture (SILAC) is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of 'heavy' 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization and quantitation of proteins.

Thermo Scientific Heavy and Light Amino Acids for SILAC are used together with specialized cell culture media that are deficient in essential amino acids. Heavy and light L-lysine and L-arginine are the most common amino acids used for SILAC analysis of tryptic peptides. Up to three different experimental conditions can be readily analyzed with different isotopes of lysine and arginine. For lysine three-plex experiments, 4,4,5,5-D4 L-lysine and 13C6 15N2 L-lysine are used to generate peptides with 4- and 8-Da mass shifts, respectively, compared to peptides generated with light lysine. For arginine three-plex experiments, 13C6 L-arginine and 13C6 15N4 L-arginine are used to generate peptides with 6- and 10-Da mass shifts, respectively, compared to peptides generated with light arginine. L-leucine is another amino acid commonly used for SILAC labeling, because it is one of the most common amino acids found in protein sequences. Proline is a non-essential amino acid that is sometimes added to SILAC media to prevent the metabolic conversion of heavy arginine to heavy proline in mammalian cell lines with high arginine dehydrogenase activity.

Related Products
L-Arginine-HCl for SILAC
L-Proline for SILAC
L-Leucine for SILAC
L-Lysine-2HCl for SILAC
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Description
L-Lysine-2HCl, 13C6, 15N2
For Use With (Equipment)
Mass Spectrometer
Product Type
Amino Acids
Label or Dye
Mass Isotope of Amino Acid
Shipping Condition
Room Temperature
Starting Material
Cells
Final Product Type
Protein
Detection Method
Mass Spectrometry
Workflow Step
Protein Labeling
Quantity
50 mg
Form
Powder

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text